Skip to Content
Merck
All Photos(1)

Key Documents

700479

Sigma-Aldrich

Ferrocenylmethyl methacrylate

95% (NMR), contains Ionol® 46 (Raschig GmbH) as inhibitor

Synonym(s):

FMMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16FeO2
CAS Number:
Molecular Weight:
284.13
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95% (NMR)

form

solid

contains

Ionol® 46 (Raschig GmbH) as inhibitor

mp

52-54 °C

storage temp.

−20°C

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.CC(=C)C(=O)OC[C]2[CH][CH][CH][CH]2

InChI

1S/C10H11O2.C5H5.Fe/c1-8(2)10(11)12-7-9-5-3-4-6-9;1-2-4-5-3-1;/h3-6H,1,7H2,2H3;1-5H;

InChI key

GSPYKNCYLXXOJO-UHFFFAOYSA-N

Legal Information

Ionol is a registered trademark of Raschig GmbH

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoonhee Na et al.
Journal of materials chemistry. B, 8(9), 1906-1913 (2020-02-12)
Ferrocene-containing nanoparticles show reversible redox activity that could trigger drug release mediated by reactive oxygen species (ROS). In this study, four ferrocene-containing polymers, comprising ferrocenylmethyl methacrylate (FMMA)-methacrylic acid (MA) random copolymers, i.e., poly(FMMA-r-MA), were synthesized via radical polymerization, resulting in
Antonio Preti et al.
Social psychiatry and psychiatric epidemiology, 54(3), 355-367 (2018-12-14)
Paranoid thoughts are relatively common in the general population and can increase the risk of developing mental health conditions. In this study, we investigate the latent structure of paranoia in a sample of young people. Cross-sectional survey; 243 undergraduate students
Liying Zhao et al.
Biosensors & bioelectronics, 150, 111895-111895 (2019-11-25)
Cytokeratin fragment antigen 21-1 (CYFRA 21-1) DNA is a crucial biomarker closely associated with non-small cell lung cancer. Here, we fabricated a novel electrochemical biosensor for ultrasensitive detection of CYFRA 21-1 DNA via polysaccharide and electrochemically mediated atom transfer radical
Ligang Ma et al.
The Analyst, 145(3), 858-864 (2019-12-18)
A novel electrochemical biosensor was reported for the first time to achieve highly sensitive DNA detection based on photoinduced atom transfer radical polymerization (photoATRP). In this work, PNA was applied as the capture probe to specifically recognize the target DNA
Yoonhee Na et al.
International journal of pharmaceutics, 596, 120205-120205 (2021-01-25)
The elevated production of reactive oxygen species (ROS) in wounded sites triggers a series of harmful effects, including cellular senescence, fibrotic scar formation, and inflammation. Therefore, alleviating oxidative stress in the microenvironment of wounded sites might promote regenerative wound healing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service