Skip to Content
Merck
All Photos(3)

Key Documents

283630

Sigma-Aldrich

Vinylferrocene

97%

Synonym(s):

Ethenylferrocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12Fe
CAS Number:
Molecular Weight:
212.07
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

core: iron
reagent type: catalyst

bp

80-85 °C/0.2 mmHg (lit.)

mp

51-53 °C (lit.)

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C=C[C]2[CH][CH][CH][CH]2

InChI

1S/C7H7.C5H5.Fe/c1-2-7-5-3-4-6-7;1-2-4-5-3-1;/h2-6H,1H2;1-5H;

InChI key

LCPVTGDQYVLJHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Synthesis of 2-aminomethylferrocenes from vinyl ferrocene and lithium amides.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takanori Tamaki et al.
The journal of physical chemistry. B, 111(34), 10312-10319 (2007-08-10)
We immobilized hydroquinone through a spacer to polymer grafted on carbon black and achieved a high-surface-area biofuel cell electrode. Quinone compounds are well-known to transfer electrons in the respiratory chain and have been considered prospective mediators in biofuel cells because
Kay L Robinson et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 24(3), 339-343 (2008-03-12)
Poly(vinylferrocene) (pVFc) homopolymer was synthesized by free radical polymerization, along with a series of pVFc-based copolymers containing either styrene, vinylanthracene or methylmethacrylate. This report details the electrochemical experiments conducted to examine the stability of the various pVFc based polymers, which
Ryoji Kurita et al.
Biosensors & bioelectronics, 20(3), 518-523 (2004-10-21)
This paper proposes a very simple procedure for preparing a biocompatible sensor based on a protein (bovine serum albumin, BSA), enzyme and vinylferrocene (VF) composite membrane modified electrode. The membrane was prepared simply by first casting vinylferrocene and then coating
Ceren Kaçar et al.
Analytical and bioanalytical chemistry, 409(11), 2873-2883 (2017-02-12)
Highly sensitive L-lysine enzyme electrodes were constructed by using poly(vinylferrocene)-multiwalled carbon nanotubes-gelatine (PVF/MWCNTs-GEL) and poly(vinylferrocene)-multiwalled carbon nanotubes-gelatine-graphene (PVF/MWCNTs-GEL/GR) composites as sensing interfaces and their performances were evaluated. Lysine oxidase (LO) was immobilized onto the composite modified glassy carbon electrodes (GCE)
Atsunori Hiratsuka et al.
Biosensors & bioelectronics, 21(6), 957-964 (2005-11-01)
We propose a new strategy for constructing a mediator-type biosensor as a Bio-MicroElectroMechanical Systems (BioMEMS) application. A vinylferrocene plasma-polymerized film (PPF) was deposited directly onto the surface of an electrode under dry conditions. The resulting redox film was extremely thin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service