Skip to Content
Merck
All Photos(2)

Key Documents

684910

Sigma-Aldrich

1H-Benzo[g]indole

97%

Synonym(s):

1H-Benz[g]indole, 6,7-Benzindole, NSC 153687

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9N
CAS Number:
Molecular Weight:
167.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

180-184 °C

SMILES string

c1ccc2c(c1)ccc3cc[nH]c23

InChI

1S/C12H9N/c1-2-4-11-9(3-1)5-6-10-7-8-13-12(10)11/h1-8,13H

InChI key

HIYWOHBEPVGIQN-UHFFFAOYSA-N

Application

  • Reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
Useful fused indole as a starter in indole chemistry.[1]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason Siu et al.
Organic & biomolecular chemistry, 2(2), 160-167 (2004-01-23)
The development of enhanced conditions for Lewis acid catalysed Leimgruber-Batcho indole synthesis using microwave acceleration is described. This approach has permitted the preparation of a variety of heteroaromatic enamine intermediates in good yield and high purities. Subsequent catalytic hydrogenation reactions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service