Skip to Content
Merck
All Photos(1)

Key Documents

03024

Sigma-Aldrich

1,1,2-Trimethylbenz[e]indole

≥98.0% (HPLC)

Synonym(s):

2,3,3-Trimethyl-4,5-benzo-3H-indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15N
CAS Number:
Molecular Weight:
209.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (HPLC)

form

crystals

mp

111-117 °C

SMILES string

CC1=Nc2ccc3ccccc3c2C1(C)C

InChI

1S/C15H15N/c1-10-15(2,3)14-12-7-5-4-6-11(12)8-9-13(14)16-10/h4-9H,1-3H3

InChI key

WJZSZXCWMATYFX-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • Fluorescent probes for imaging of tumors in vivo
  • Hypoxia-sensitive fluorescent probes for in vivo real-time imaging of acute ischemia
  • Near IR fluorescent probe for imaging integrin receptor expression

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Song-Jiao Li et al.
Talanta, 188, 691-700 (2018-07-22)
Carbon monoxide is a critical gasotransmitter in the body and related with mitochondrial respiration. To date, various fluorescent probes for CO have been well proposed, but two main problems remain. One is that most of the probes are not mitochondria-targeting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service