Skip to Content
Merck
All Photos(1)

Key Documents

680788

Sigma-Aldrich

5-(3-Pyridyl)-1,3,4-oxadiazole-2-thiol

97%

Synonym(s):

5-(3-Pyridinyl)-1,3,4-oxadiazole-2(3H)-thione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N3OS
CAS Number:
Molecular Weight:
179.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
97%

Quality Level

Assay

97%

form

solid

mp

231 °C (dec.)

SMILES string

Sc1nnc(o1)-c2cccnc2

InChI

1S/C7H5N3OS/c12-7-10-9-6(11-7)5-2-1-3-8-4-5/h1-4H,(H,10,12)

InChI key

QBLWWQDTKCIRSW-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service