67873
4-Methylmorpholine N-oxide monohydrate
≥95.0% (N)
Synonym(s):
4-Methylmorpholine 4-oxide monohydrate, 4-Methylmorpholine oxide monohydrate, N-Methylmorpholine N-oxide monohydrate, N-Methylmorpholine oxide monohydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H11NO2 · H2O
CAS Number:
Molecular Weight:
135.16
Beilstein:
507437
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95.0% (N)
mp
71-75 °C
functional group
ether
SMILES string
O.C[N+]1([O-])CCOCC1
InChI
1S/C5H11NO2.H2O/c1-6(7)2-4-8-5-3-6;/h2-5H2,1H3;1H2
InChI key
WAZPLXZGZWWXDQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
4-Methylmorpholine N-oxide (MNO) monohydrate can be used:
- As a reagent in the dihydroxylation and aminohydroxylation of olefins in the presence of osmium tetroxide (OsO4) catalyst.
- In the selective oxidation of sulphides to sulfoxides or sulfones in the presence of manganese and copper complexes with tetradentate ligands.
Other Notes
Reagent for an improved catalytic OsO4 oxidation of olefins to cis-1,2-diols; Review; For a ruthenium catalysed oxidation of alcohols to aldehydes and ketones
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A New Approach to Osmium-Catalyzed Asymmetric Dihydroxylation and Aminohydroxylation of Olefins
Andersson MA, et al.
Angewandte Chemie (International Edition in English), 41(3), 472-475 (2002)
V. Van Rheenen et al.
Tetrahedron Letters, 1973-1973 (1976)
K.B. Sharpless et al.
Tetrahedron Letters, 2503-2503 (1976)
Homogeneous and encapsulated within the cavities of zeolite Y chiral manganese and copper complexes with C2-multidentate ligands as catalysts for the selective oxidation of sulphides to sulfoxides or sulfones
Alcon MJ, et al.
J. Mol. Catal. A: Chem., 178(1-2), 253-266 (2002)
M. Schroder
Chemical Reviews, 80, 187-187 (1980)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service