Skip to Content
Merck
All Photos(3)

Key Documents

M56557

Sigma-Aldrich

4-Methylmorpholine

ReagentPlus®, 99%

Synonym(s):

N-Methylmorpholine, NMM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO
CAS Number:
Molecular Weight:
101.15
Beilstein:
102719
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
bp:
115-116 °C/750 mmHg (lit.)
vapor pressure:
18 mmHg ( 20 °C)

vapor density

>1 (vs air)

Quality Level

vapor pressure

18 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

dilution

(for general lab use)

refractive index

n20/D 1.435 (lit.)

bp

115-116 °C/750 mmHg (lit.)

mp

−66 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

SMILES string

CN1CCOCC1

InChI

1S/C5H11NO/c1-6-2-4-7-5-3-6/h2-5H2,1H3

InChI key

SJRJJKPEHAURKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-methylmorpholine, also known as N-Methylmorpholine, is a cyclic tertiary amine and is commonly used as a highly valuable solvent and a fine chemical for cellulose.

Application

4-Methylmorpholine can be used to synthesize N-butyl-N-methylmorpholinium bromide by reacting with bromobutane.
4-methylmorpholine is used:
  • in the synthesis of N-methylmorpholine-N-oxide (NMMO)
  • as an effective and inexpensive additive for highly enantioselective arylation of aryl aldehydes catalyzed by the (S)-H8-BINOL-Ti(Oi-Pr)2 complex

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup - DIN 51755 Part 1

Flash Point(C)

12 °C - closed cup - DIN 51755 Part 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benedetta Del Secco et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 18(9), 2142-2149 (2019-04-24)
Silica nanoparticles (NPs) are versatile nanomaterials, which are safe with respect to biomedical applications, and therefore are highly investigated. The advantages of NPs include their ease of preparation, inexpensive starting materials and the possibility of functionalization or loading with various
Size-controlled electrochemical synthesis of palladium nanoparticles using morpholinium ionic liquid.
Cha J H, et al.
Korean Journal of Chemical Engineering, 24(6), 1089-1094 (2007)
V Balachandran et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 97, 1101-1110 (2012-08-30)
The experimental FT-IR (4000-400 cm(-1)) and FT-Raman (3500-100 cm(-1)) spectra of 4-Methylmorpholine were recorded. The observed bands were interpreted with the aid of normal coordinate analysis and force field calculations based on density functional theory (DFT) using B3LYP functional theory
L Hansén et al.
Scandinavian journal of work, environment & health, 12(1), 66-69 (1986-02-01)
The N-methylmorpholine levels in workroom air in a polyurethane foam factory were determined by two methods in which midget impinger flasks were used for sampling. The analyses were performed by gas chromatography and isotachophoresis. The values obtained by the gas
Sophie C Larnaudie et al.
Biomaterials, 178, 570-582 (2018-04-24)
Size and shape have progressively appeared as some of the key factors influencing the properties of nanosized drug delivery systems. In particular, elongated materials are thought to interact differently with cells and therefore may allow alterations of in vivo fate without

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service