Skip to Content
Merck
All Photos(2)

Key Documents

652458

Sigma-Aldrich

Barium fluoride

precipitated, 99.95% trace metals basis

Synonym(s):

Barium difluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaF2
CAS Number:
Molecular Weight:
175.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Quality Level

Assay

99.95% trace metals basis

form

powder

quality

precipitated

impurities

≤550.0 ppm Trace Metal Analysis

bp

2260 °C (lit.)

density

4.830 g/mL at 25 °C
4.89 g/mL at 25 °C (lit.)

SMILES string

F[Ba]F

InChI

1S/Ba.2FH/h;2*1H/q+2;;/p-2

InChI key

OYLGJCQECKOTOL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Primary glass modifier in fluorozirconate glasses.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dean C Sayle et al.
Physical chemistry chemical physics : PCCP, 7(1), 16-18 (2005-01-07)
We show, using simulation techniques, that the high ionic conductivity in BaF2/CaF2 heterolayers is because the interfaces reduce the activation energy barriers to mobility and increase the number of charge carriers.
Yongqian Lei et al.
Dalton transactions (Cambridge, England : 2003), 40(1), 142-145 (2010-11-16)
Hydrophilic Ce, Tb doped BaYF(5) nanocrystals with uniform size were synthesized by a microwave-assisted route. The synthesized nanocrystals can be well dispersed in hydrophilic solutions (DMSO, DMF, EG, H(2)O). This synthesis procedure represents a less time consuming method, with high
Jian Ruan et al.
Optics express, 17(7), 5163-5169 (2009-04-01)
Bi-doped BaF(2) crystal was grown by the temperature gradient technique and its spectral properties were investigated. The absorption, emission and excitation spectra were measured at room temperature. Two broadband emissions centered at 1070 and 1500 nm were observed in Bi-doped
S Tavernier et al.
Medical progress through technology, 17(3-4), 237-241 (1991-01-01)
The work presented is part of a design study for a Positron Emission Tomograph (PET) scanner based on the use of BaF2 scintillator and photosensitive wire chambers. The detection efficiency for gamma radiation of 511 keV is found close to
H Nanto et al.
Radiation protection dosimetry, 100(1-4), 385-388 (2002-10-18)
BaFI:Eu phosphors are fabricated using a new method of synthesis: liquid phase synthesis, in which the phosphor particles are formed through the association of Ba2+ ions, F-ions and Eu2+ ions in solution. An intense optically stimulated luminescence (OSL) peak at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service