Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Barium fluoride
Synonym(s):
Barium fluoride
Empirical Formula (Hill Notation):
BaF
2
CAS Number:
7787-32-8
Molecular Weight:
175.32
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (3)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
202746
Barium fluoride,
99.99% trace metals basis
Expand
View Pricing
236101
Barium fluoride,
powder, 98%
Expand
View Pricing
652458
Barium fluoride,
precipitated, 99.95% trace metals basis
Expand
View Pricing
1.01722
Barium fluoride,
99.99 Suprapur
®
Expand
View Pricing