Skip to Content
Merck
All Photos(4)

Key Documents

637017

Sigma-Aldrich

Bismuth(III) oxide

nanopowder, 90-210 nm particle size, 99.8% trace metals basis

Synonym(s):

Dibismuth trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi2O3
CAS Number:
Molecular Weight:
465.96
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

description

beta

Quality Level

Assay

99.8% trace metals basis

form

nanopowder
spherical

reaction suitability

reagent type: catalyst
core: bismuth

surface area

3.2-3.5 m2/g

particle size

90-210 nm

bulk density

0.5‑1.1 g/mL

SMILES string

O=[Bi]O[Bi]=O

InChI

1S/2Bi.3O

InChI key

WMWLMWRWZQELOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Characterization and Physical Properties of Nano-sized Bi2O3/Polymer: This research investigates the properties of bismuth(III) oxide nanoparticles within a polymer matrix, emphasizing energy and high-refractive index applications. It explores the potential of Bi2O3 in enhancing the performance of composite materials (A Alruwaili, AM El Sayed, 2024).
  • Electroanalytical Performance Enhancement by Bismuth (III) Oxide: A study on incorporating bismuth(III) oxide nanoparticles into carbon ceramic electrodes to improve their electroanalytical capabilities, particularly for detecting pollutants in water (M Brycht et al., 2024).
  • Visible Light Responsive Photocatalysts for Arsenic Remediation: This review highlights the role of bismuth-based photocatalysts, including bismuth(III) oxide, in the effective remediation of arsenic from water sources, underlining the photocatalytic capabilities of these materials (I Natali Sora et al., 2024).
  • High-Performance Composites for Gamma-Ray Shielding: An investigation into fluoroelastomer composites filled with bismuth(III) oxide for their application in gamma-ray shielding, highlighting the material′s effectiveness in blocking radiation (SM Magnere et al., 2024).

Features and Benefits

Preparation of new stabilized, oxide ion-conducting, bismuth vanadate phases by a microwave assisted method, from V2O5, Bi2O3 and other solid oxides, was reported. These ceramics show promise in solid oxide fuel cells, water-vapor electrolyzers and oxygen sensors.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu You et al.
ACS nano, 6(6), 5388-5394 (2012-05-18)
The novel strain-driven morphotropic phase boundary (MPB) in highly strained BiFeO(3) thin films is characterized by well-ordered mixed phase nanodomains (MPNs). Through scanning probe microscopy and synchrotron X-ray diffraction, eight structural variants of the MPNs are identified. Detailed polarization configurations
Rifath Sharmin et al.
Journal of the Air & Waste Management Association (1995), 62(9), 1032-1039 (2012-10-02)
Photocatalytic oxidation (PCO) is a promising technology for indoor air purification due to low operating cost, potentially long service life, and low maintenance. Ultraviolet light-emitting diode (UVLED) is a new concept in the field of PCO, which has several advantages
Marco Antonio Hungaro Duarte et al.
Journal of endodontics, 38(3), 394-397 (2012-02-22)
The aim of this study was to evaluate the pH, calcium ion release, setting time, and solubility of white mineral trioxide aggregate (WMTA) and white Portland cement (WPC) combined with the following radiopacifying agents: bismuth oxide (BO), calcium tungstate (CT)
V S Kopp et al.
Acta crystallographica. Section A, Foundations of crystallography, 68(Pt 1), 148-155 (2011-12-22)
X-ray diffraction from films consisting of layers with different thicknesses, structures and chemical contents is analysed. The disorder is described by probabilities for different sequences of layers. Closed analytical expressions for the diffracted X-ray intensity are obtained when the layers
L M Formosa et al.
Dental materials : official publication of the Academy of Dental Materials, 28(5), 584-595 (2012-03-14)
Tricalcium silicate is the major constituent phase in mineral trioxide aggregate (MTA). It is thus postulated that pure tricalcium silicate can replace the Portland cement component of MTA. The aim of this research was to evaluate the microstructure and surface

Articles

Innovation in dental restorative materials is driven by the need for biocompatible and natural-appearing restoration alternatives. Conventional dental materials like amalgam and composite resins have inherent disadvantages.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service