Skip to Content
Merck
All Photos(1)

Key Documents

449865

Sigma-Aldrich

Lead(II) chloride

AnhydroBeads, −10 mesh, 99.999%

Synonym(s):

Cotunnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbCl2
CAS Number:
Molecular Weight:
278.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999%

vapor pressure

1 mmHg ( 547 °C)

Quality Level

product line

AnhydroBeads

Assay

99.999%

reaction suitability

reagent type: catalyst
core: lead

impurities

≤15.0 ppm Trace Metal Analysis

particle size

−10 mesh

bp

950 °C (lit.)

mp

501 °C (lit.)

density

5.85 g/mL at 25 °C (lit.)

SMILES string

Cl[PbH2]Cl

InChI

1S/2ClH.Pb/h2*1H;/q;;+2/p-2

InChI key

HWSZZLVAJGOAAY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1A - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Okada et al.
Journal of environmental management, 110, 207-214 (2012-07-14)
This study investigated the effect of the chemical composition of incineration fly ash on the leaching characteristics of Pb from melting furnace fly ash generated by melting incineration fly ash. Melting furnace fly ash from both a real-scale melting process
Muhammad Rafique et al.
Journal of the College of Physicians and Surgeons--Pakistan : JCPSP, 19(8), 510-513 (2009-08-05)
To determine the effects of lead and zinc administration on the quality of semen of albino rats. Experimental study. Basic Medical Sciences Institute, Jinnah Postgraduate Medical Centre, Karachi, from August 2003 to December 2005. Sixty adult albino rats selected for
Iris Zeller et al.
Arteriosclerosis, thrombosis, and vascular biology, 30(9), 1733-1740 (2010-07-03)
To validate the hypothesis that the toxic heavy metal lead (Pb) may be linked to cardiovascular diseases via the initiation of atherosclerosis, in vivo and in vitro studies were conducted. During the human study part of this project, serum Pb
Md Kawser Ahmed et al.
Environmental monitoring and assessment, 182(1-4), 197-204 (2011-01-07)
The present investigation was undertaken to study the induction of DNA damage by lead chloride (PbCl(2)) in freshwater climbing perch Anabas testudineus using alkaline single cell gel electrophoresis (comet assay). Based on the LC(50) values of lead chloride of A.
Sun Li et al.
Toxicology in vitro : an international journal published in association with BIBRA, 22(8), 1815-1819 (2008-09-16)
Lead exposure is one of major public health problems. The aim of this study was to examine in vitro the effect of lead on lymphocyte intracellular Ca(2+) ([Ca(2+)]i). Lymphocytes were separated from mice spleen and treated with three doses of

Articles

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service