Skip to Content
Merck
All Photos(1)

Key Documents

34032

Supelco

Aflatoxin G1 solution

2 μg/mL in acetonitrile, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12O7
CAS Number:
Molecular Weight:
328.27
Beilstein:
1299768
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

2 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

COc1cc2OC3OC=CC3c2c4OC(=O)C5=C(CCOC5=O)c14

InChI

1S/C17H12O7/c1-20-9-6-10-12(8-3-5-22-17(8)23-10)14-11(9)7-2-4-21-15(18)13(7)16(19)24-14/h3,5-6,8,17H,2,4H2,1H3

InChI key

XWIYFDMXXLINPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lokman Alpsoy et al.
Human & experimental toxicology, 30(6), 515-519 (2010-07-16)
Aflatoxins have been shown to be hepatotoxic, carcinogenic, mutagenic and teratogenic to different species of animals. Besides, at low concentrations, Selenium (Se(4+)) is antimutagenic and anticarcinogenic while it is toxic, mutagenic and carcinogenic at high concentrations. In this study, we
Haitao Shen et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(9), 3222-3228 (2012-06-20)
Our previous studies showed intragastric administration of Aflatoxin G(1) (AFG(1)) could induce lung adenocarcinoma, which derived from alveolar type II cells (AT-II cells). AT-II cells contribute to Aflatoxin B(1) (AFB(1)) metabolism and also serve as the potential progenitor cells for
Magda M Aly et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48(11), 3031-3034 (2010-05-18)
Different local and exported white cheese samples were collected from different markets in Jeddah during September 2008. Trace and heavy metals including Pb, Zn, Mn, Cu, Fe and Cd were analyzed using atomic absorption spectrometry. The concentration of the tested
Hongmei Zeng et al.
Applied microbiology and biotechnology, 90(2), 635-650 (2010-12-15)
In aflatoxin biosynthesis, aflatoxins G(1) (AFG(1)) and B(1) (AFB(1)) are independently produced from a common precursor, O-methylsterigmatocystin (OMST). Recently, 11-hydroxy-O-methylsterigmatocystin (HOMST) was suggested to be a later precursor involved in the conversion of OMST to AFB(1), and conversion of HOMST
Zengning Li et al.
Wei sheng yan jiu = Journal of hygiene research, 40(1), 50-52 (2011-03-26)
To explore the effect of AFG1 on the expression of human leukocyte antigen (HLA-I) molecule in human esophageal epithelial cells. Western Blot and RT-PCT analysis was used to explore the effect of AFG1 on the antigen presenting function of primary

Protocols

LC/MS/MS Analysis of Aflatoxins in Hops on Ascentis® Express 2.7 μm Phenyl-Hexyl after Cleanup Using Supel™ Tox AflaZea SPE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service