596566
Calcium
dendritic pieces, purified by distillation, 99.9% trace metals basis
Synonym(s):
Calcium atom, Calcium element
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Ca
CAS Number:
Molecular Weight:
40.08
Beilstein:
4241647
EC Number:
MDL number:
UNSPSC Code:
12141503
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
99.9% trace metals basis
form
dendritic pieces
purified by
distillation
reaction suitability
reagent type: reductant
resistivity
3.5 μΩ-cm, 20°C
bp
1484 °C (lit.)
mp
850 °C (lit.)
density
1.54 g/mL at 25 °C (lit.)
SMILES string
[Ca]
InChI
1S/Ca
InChI key
OYPRJOBELJOOCE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Precursor to Ca8Al3 and Ca13Al14; intermetallics with new metallic Zintl phases.7
accessory
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Water-react 2
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tsai-Wen Chen et al.
Nature, 499(7458), 295-300 (2013-07-23)
Fluorescent calcium sensors are widely used to image neural activity. Using structure-based mutagenesis and neuron-based screening, we developed a family of ultrasensitive protein calcium sensors (GCaMP6) that outperformed other sensors in cultured neurons and in zebrafish, flies and mice in
S Boonen et al.
Journal of internal medicine, 259(6), 539-552 (2006-05-18)
Combined calcium and vitamin D supplementation is an essential component of the management of osteoporosis, supported by a strong scientific rationale. The types of individuals who should receive calcium and vitamin D supplements are those: (i) patients with documented osteoporosis
Byrappa Venkatesh et al.
Nature, 505(7482), 174-179 (2014-01-10)
The emergence of jawed vertebrates (gnathostomes) from jawless vertebrates was accompanied by major morphological and physiological innovations, such as hinged jaws, paired fins and immunoglobulin-based adaptive immunity. Gnathostomes subsequently diverged into two groups, the cartilaginous fishes and the bony vertebrates.
Eleni Gourni et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(10), 1719-1725 (2014-08-26)
Gastrin-releasing peptide receptors (GRPrs) are overexpressed on a variety of human cancers, providing the opportunity for peptide receptor targeting via radiolabeled bombesin-based peptides. As part of our ongoing investigations into the development of improved GRPr antagonists, this study aimed at
Virginia Nguyen et al.
Heart (British Cardiac Society), 101(12), 943-947 (2015-02-07)
Aortic valve stenosis (AS) is a progressive disease, but the impact of baseline AS haemodynamic or anatomic severity on AS progression remains unclear. In 149 patients (104 mild AS, 36 moderate AS and 9 severe AS) enrolled in 2 ongoing
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service