Skip to Content
Merck
All Photos(3)

Key Documents

596388

Sigma-Aldrich

Formaldehyde-13C, d2 solution

20 wt. % in D2O, ≥99 atom % 13C, ≥98 atom % D

Synonym(s):

13C and deuterium labeled formaldehyde, Formalin-13C,d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D13CDO
CAS Number:
Molecular Weight:
33.03
MDL number:
UNSPSC Code:
12352501
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥99 atom % 13C
≥98 atom % D

Quality Level

concentration

20 wt. % in D2O

technique(s)

protein expression: suitable

mass shift

M+3

SMILES string

[2H][13C]([2H])=O

InChI

1S/CH2O/c1-2/h1H2/i1+1D2

InChI key

WSFSSNUMVMOOMR-WGVGGRBOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Plum et al.
Immunity, 52(2), 404-416 (2020-02-13)
Mast cells are rare tissue-resident cells of importance to human allergies. To understand the structural basis of principle mast cell functions, we analyzed the proteome of primary human and mouse mast cells by quantitative mass spectrometry. We identified a mast-cell-specific
Yi Li et al.
STAR protocols, 1(3), 100137-100137 (2020-12-31)
Alk-Ph is a clickable APEX2 substrate developed for spatially restricted protein/RNA labeling in intact yeast cells. Alk-Ph is more water soluble and cell wall permeable than biotin-phenol substrate, allowing more efficient profiling of the subcellular proteome in microorganisms. We describe
Man Pan et al.
Nature communications, 12(1), 121-121 (2021-01-07)
p97, also known as valosin-containing protein (VCP) or Cdc48, plays a central role in cellular protein homeostasis. Human p97 mutations are associated with several neurodegenerative diseases. Targeting p97 and its cofactors is a strategy for cancer drug development. Despite significant
Jie Wang et al.
Nature, 569(7757), 509-513 (2019-05-10)
A universal gain-of-function approach for selective and temporal control of protein activity in living systems is crucial to understanding dynamic cellular processes. Here we report development of a computationally aided and genetically encoded proximal decaging (hereafter, CAGE-prox) strategy that enables time-resolved
Victoria Garrido et al.
Molecular systems biology, 17(10), e10145-e10145 (2021-10-07)
Bacteria present a promising delivery system for treating human diseases. Here, we engineered the genome-reduced human lung pathogen Mycoplasma pneumoniae as a live biotherapeutic to treat biofilm-associated bacterial infections. This strain has a unique genetic code, which hinders gene transfer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service