Skip to Content
Merck
All Photos(2)

Key Documents

593648

Sigma-Aldrich

Phytic acid solution

50 % (w/w) in H2O

Synonym(s):

myo-Inositol hexakis(dihydrogen phosphate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H18O24P6
CAS Number:
Molecular Weight:
660.04
Beilstein:
2201952
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

45.0-55.0 %

concentration

50 % (w/w) in H2O

bp

105 °C

solubility

water: soluble at 20 °C

density

1.432 g/mL at 25 °C

functional group

phosphate

SMILES string

OP(O)(=O)O[C@@H]1[C@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@H]1OP(O)(O)=O

InChI

1S/C6H18O24P6/c7-31(8,9)25-1-2(26-32(10,11)12)4(28-34(16,17)18)6(30-36(22,23)24)5(29-35(19,20)21)3(1)27-33(13,14)15/h1-6H,(H2,7,8,9)(H2,10,11,12)(H2,13,14,15)(H2,16,17,18)(H2,19,20,21)(H2,22,23,24)/t1-,2-,3-,4+,5-,6-

InChI key

IMQLKJBTEOYOSI-GPIVLXJGSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phytic acid is a mineral chelator that can bind to minerals to form mineral-phytate complex.It can also complex with metal ions bound to the surface of magnesium alloy to form a conversion coating, which can improve the resistance of magnesium alloy towards corrosion.

Application

  • Phytic acid in dental applications: A 2024 study explores the effectiveness of phytic acid as an irrigation solution to improve the adhesion and sealing properties of calcium silicate-based cements in endodontic treatments, demonstrating its potential as a chelating agent in dental procedures (Koçak Şahin and Ünal, 2024).
  • Phytic acid in energy storage: Phytic acid is utilized to stabilize magnesium metal anodes in battery technology, contributing to the development of more durable and efficient energy storage systems (Wen et al., 2024).
  • Phytic acid in battery technology: Another 2024 study incorporates phytic acid in the development of a hydrogel polymer electrolyte and cathode material for rechargeable zinc metal hydrogel batteries, highlighting its role in enhancing the performance and stability of battery systems (Dilwale et al., 2024).
  • Phytic acid in strain sensors: Research focuses on the use of a nacre-mimetic structure multifunctional ion-conductive hydrogel, enhanced by phytic acid, for developing high-performance strain sensors with significant implications in wearable technology and robotics (Wang et al., 2024).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Study on the corrosion resistance of phytic acid conversion coating for magnesium alloys"
Jianrui L, et al.
Surface and Coatings Technology, 201(03), 1536- 1541 (2006)
"The effect of minerals and mineral chelators on the formation of phytase-resistant and phytase-susceptible forms of phytic acid in solution and in a slurry of canola meal"
Maenz.DD, et al.
Anim. Feed Sci. Technol., 81(03), 177-192 (1999)
Xiangdong Long et al.
Nature communications, 11(1), 4074-4074 (2020-08-15)
Single-atom metal-nitrogen-carbon (M-N-C) catalysts have sparked intensive interests, however, the development of an atomically dispersed metal-phosphorus-carbon (M-P-C) catalyst has not been achieved, although molecular metal-phosphine complexes have found tremendous applications in homogeneous catalysis. Herein, we successfully construct graphitic phosphorus species
Jorge Escorihuela et al.
Polymers, 12(6) (2020-06-24)
The use of phosphoric acid doped polybenzimidazole (PBI) membranes for fuel cell applications has been extensively studied in the past decades. In this article, we present a systematic study of the physicochemical properties and proton conductivity of PBI membranes doped
Ines Boukhris et al.
PloS one, 15(4), e0231397-e0231397 (2020-04-18)
The increasing challenge of antibiotic resistance requires not only the discovery of new antibiotics, but also the development of new alternative approaches. Therefore, in the present study, we investigated for the first time the antibacterial potential of phytic acid (myo-inositol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service