Skip to Content
Merck
All Photos(1)

Key Documents

542229

Sigma-Aldrich

2-Chloro-3′,4′-dimethoxybenzil

97%

Synonym(s):

1-(2-Chlorophenyl)-2-(3,4-dimethoxyphenyl)ethane-1,2-dione

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4(CO)2C6H3(OCH3)2
CAS Number:
Molecular Weight:
304.73
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

119-123 °C (lit.)

SMILES string

COc1ccc(cc1OC)C(=O)C(=O)c2ccccc2Cl

InChI

1S/C16H13ClO4/c1-20-13-8-7-10(9-14(13)21-2)15(18)16(19)11-5-3-4-6-12(11)17/h3-9H,1-2H3

InChI key

ULVSCSFZMZRZHJ-UHFFFAOYSA-N

Gene Information

General description

2-Chloro-3′,4′-dimethoxybenzil (CDMB, o-chlorobenzveratroin) is a specific inhibitor of human carboxylesterase-2 (hCE-2).

Application

2-Chloro-3′,4′-dimethoxybenzil (2-Chloro-3,4-dimethoxybenzil) may be used in the synthesis of 2-(2-chlorophenyl)-3-(3,4-dimethoxyphenyl)quinoxaline and 3-(2-chlorophenyl)-4-(3,4-dimethoxyphenyl)-2,5-diphenylcyclopenta-2,4-dienone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quan-Gang Zhu et al.
Biological & pharmaceutical bulletin, 30(3), 532-536 (2007-03-03)
There is increasing evidence that epidermal carboxylesterase may be involved in the stereoselective hydrolysis of prodrugs in percutaneous absorption. The present study was designed to evaluate the stereoselective characteristics and mechanisms of ketoprofen ethyl ester hydrolysis by epidermal carboxylesterase expressed
Microwave-Assisted Synthesis of Functionalized Shvo-Type Complexes.
Cesari C, et al.
Organometallics, 33(11), 2814-2819 (2014)
2-(2-Chlorophenyl)-3-(3, 4-dimethoxyphenyl) quinoxaline.
Cantalupo SA, et al.
Acta Crystallographica Section E, Structure Reports Online, 66(8), o2184-o2184 (2010)
Mixed benzoins. Ix. Meso chloro derivatives.
Buck JS and Ide WS.
Journal of the American Chemical Society, 54(11), 4359-4365 (1932)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service