Skip to Content
Merck
All Photos(3)

Documents

538035

Sigma-Aldrich

1,4-Cyclohexanedicarboxylic acid

99%

Synonym(s):

1,4-CHDA-HP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(CO2H)2
CAS Number:
Molecular Weight:
172.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

164-167 °C (lit.)

SMILES string

OC(=O)C1CCC(CC1)C(O)=O

InChI

1S/C8H12O4/c9-7(10)5-1-2-6(4-3-5)8(11)12/h5-6H,1-4H2,(H,9,10)(H,11,12)

InChI key

PXGZQGDTEZPERC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Cyclohexanedicarboxylic acid (1,4-CHDA) has cyclohexane based structure. CHDA has better weatherability, higher impact strength, and faster stress relaxation. CHDA exhibits three isomeric forms:
  • axial, axial (a,a) trans-isomer
  • equatorial, equatorial (e,e) trans-isomer
  • a,e cis-isomer

Application

1,4-Cyclohexanedicarboxylic acid may be used in the synthesis of:
  • polyesters
  • polyamides
  • poly(butylene adipate-co-butylene 1,4-cyclohexanedicarboxylate) copolyester
  • various polyester polyols

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advanced Polymerization and Properties of Biobased High T g polyester of Isosorbide and 1, 4-Cyclohexanedicarboxylic Acid through in Situ Acetylation.
Yoon WJ, et al.
Macromolecules, 46(8), 2930-2940 (2013)
Controlled stereochemistry of polyamides derived from cis/trans-1, 4-cyclohexanedicarboxylic acid.
Vanhaecht B, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 39(6), 833-840 (2001)
Synthesis and characterization of transparent and high impact resistance polyurethane coatings based on polyester polyols and isocyanate trimers.
Zhang J, et al.
Progress in Organic Coatings, 75(4), 579-583 (2012)
Role of cis-1, 4-cyclohexanedicarboxylic acid in the regulation of the structure and properties of a poly (butylene adipate-co-butylene 1, 4-cyclohexanedicarboxylate) copolymer.
Liu F, et al.
Royal Society of Chemistry Advances, 6(70), 65889-65897 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service