Skip to Content
Merck
All Photos(3)

Key Documents

123463

Sigma-Aldrich

1,2-Cyclohexanedicarboxylic anhydride, predominantly cis

95%

Synonym(s):

Hexahydrophthalic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O3
CAS Number:
Molecular Weight:
154.16
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

bp

158 °C/17 mmHg (lit.)

mp

32-34 °C (lit.)

SMILES string

O=C1OC(=O)[C@H]2CCCC[C@@H]12

InChI

1S/C8H10O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h5-6H,1-4H2/t5-,6+

InChI key

MUTGBJKUEZFXGO-OLQVQODUSA-N

General description

Predominantly cis 1,2-cyclohexanedicarboxylic anhydride (HHPA) is a cyclic anhydride that can be used for a variety of applications such as: plasticizer, rust inhibitor, and a curing agent for epoxy based resins.

Application

HPPA, in combination with triethaylamine (TEA), can be used as a polymerization initiator in the preparation of polyester based resins. It can also be used as a hardener to cure 1,4-butanediol diglycidyl ether which can be used as an epoxy based system for electronic devices.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gunvor Johannesson et al.
Toxicology and applied pharmacology, 194(1), 69-78 (2004-01-20)
Hexahydrophthalic anhydride (HHPA), an industrially important chemical, is a highly allergenic compound. The aim of this work was to identify proteins in nasal lavage fluid (NLF) that form adducts with HHPA. Such bindings may induce production of specific immunoglobulin E
Gunvor A Johannesson et al.
Biomedical chromatography : BMC, 22(3), 327-332 (2007-10-17)
An immunoaffinity extraction (IAE) column was prepared for extraction of adducts between human serum albumin (HSA) and hexahydrophthalic anhydride (HHPA). HHPA is a strong sensitizer inducing immunoglobulin E antibodies in vivo. Polyclonal antibodies from a rabbit immunized with keyhole limpet
J Nielsen et al.
Scandinavian journal of work, environment & health, 27(5), 327-334 (2001-11-20)
This study clarified the exposure-response relationships for the organic acid anhydrides (OAA) hexahydrophthalic (HHPA) and methylhexahydrophthalic (MHHPA) anhydrides and the development of specific immunoglobulin (IG) E and G antibodies and work-related symptoms. In an epoxy resin-using factory, air levels of
Monica H Kristiansson et al.
Rapid communications in mass spectrometry : RCM, 18(14), 1592-1598 (2004-07-30)
Organic acid anhydrides (OAAs) are low molecular weight, reactive compounds extensively used in industry. Exposure to these compounds may lead to allergic symptoms such as rhinitis and asthma. It is important to develop better and more informative methods for assessment
David Virley et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 24(1), 24-41 (2003-12-23)
The purpose of the present set of studies was to develop a new primate model of focal ischemia with reperfusion for long-term functional assessment in the common marmoset. Initially, the cerebral vascular anatomy of the marmoset was interrogated by Araldite-cast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service