Skip to Content
Merck
All Photos(1)

Key Documents

531634

Sigma-Aldrich

Bis(pyridine)iodonium tetrafluoroborate

Synonym(s):

Barluenga reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10BF4IN2
CAS Number:
Molecular Weight:
371.91
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reagent type: oxidant

mp

137-141 °C (lit.)

storage temp.

2-8°C

SMILES string

[I+].F[B-](F)(F)F.c1ccncc1.c2ccncc2

InChI

1S/2C5H5N.BF4.I/c2*1-2-4-6-5-3-1;2-1(3,4)5;/h2*1-5H;;/q;;-1;+1

InChI key

JBVUIHBKNVHCKK-UHFFFAOYSA-N

General description

Bis(pyridine)iodonium Tetrafluoroborate (Barluenga′s reagent) is a mild iodinating and oxidizing reagent capable of selectively reacting with a wide range of unsaturated substrates and tolerates a variety of functional groups.

Application

Bis(pyridine)iodonium Tetrafluoroborate reacts with acetonides derived from simple terpenes to accomplish selective iodofunctionalization with excellent regio- and diastereofacial control. It has been used as a reactant involved in:

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total diastereofacial selective iodofunctionalization of terpene derivatives based on Ipy2BF4
Jose Barluenga et al.
The Journal of Organic Chemistry, 68(17), 6583-6586 (2003)
?-Iodination of enaminones with bis(pyridine)iodonium(I) tetrafluoroborate
Campos, P. J. et al
Tetrahedron Letters, 38(48), 8397-8400 (1997)
José Barluenga et al.
The Journal of organic chemistry, 68(17), 6583-6586 (2003-08-16)
Acetonides 1, easily obtained from simple terpenes, react with bispyridine iodonium (I) tetrafluoroborate (Ipy(2)BF(4)) and tetrafluoroboric acid in the presence of nucleophiles to give the corresponding adducts 2 with complete regio and diastereofacial control. Acetonides 1 containing a properly located
José Barluenga et al.
Organic letters, 5(22), 4121-4123 (2003-10-24)
[reaction: see text]. Iodonium ions, o-alkynyl-substituted carbonyl compounds, and alkynes react to give 1-iodonaphthalene derivatives featuring interesting substitution patterns. The reaction with alkenes instead of acetylenes affords related naphthyl ketone derivatives. These two metal-free processes are conducted at room temperature
Regioselective synthesis of substituted naphthalenes: a novel de novo approach based on a metal-free protocol for stepwise cycloaddition of o-alkynylbenzaldehyde derivatives with either alkynes or alkenes
Jose Barluenga et al.
Organic Letters, 5(22), 4121-4123 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service