Skip to Content
Merck
All Photos(1)

Key Documents

52730

Sigma-Aldrich

Hexamethylphosphoramide

purum, ≥98.0% (GC)

Synonym(s):

HMPA, Hexamethylphosphoric acid triamide, Tris(dimethylamino)phosphine oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]3PO
CAS Number:
Molecular Weight:
179.20
Beilstein:
1099903
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.18 (vs air)

Quality Level

vapor pressure

0.07 mmHg ( 25 °C)

grade

purum

Assay

≥98.0% (GC)

impurities

≤2.0% water (Karl Fischer)

refractive index

n20/D 1.459 (lit.)
n20/D 1.459

bp

230-232 °C/740 mmHg (lit.)

mp

7 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

functional group

phosphoramide

SMILES string

CN(C)P(=O)(N(C)C)N(C)C

InChI

1S/C6H18N3OP/c1-7(2)11(10,8(3)4)9(5)6/h1-6H3

InChI key

GNOIPBMMFNIUFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Dam. 1 - Muta. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

291.2 °F - closed cup

Flash Point(C)

144 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Knud Nairz et al.
Genome biology, 5(10), R83-R83 (2004-10-06)
We report the use of the cross-linking drug hexamethylphosphoramide (HMPA), which introduces small deletions, as a mutagen suitable for reverse genetics in the model organism Drosophila melanogaster. A compatible mutation-detection method based on resolution of PCR fragment-length polymorphisms on standard
Robert B Grossman et al.
The Journal of organic chemistry, 68(3), 871-874 (2003-02-01)
Hexamethylphosphorous triamide (HMPT) and other phosphoramidites and phosphites have been found to be efficient catalysts for the Michael reaction of alkenones and alkynones with malonates, alpha-cyano esters, beta-keto esters, and nitro compounds. The relatively nontoxic, easily hydrolyzed HMPT catalyzes the
Drug-HPMA-HuIg conjugates effective against human solid cancer.
Blanka Ríhová et al.
Advances in experimental medicine and biology, 519, 125-143 (2003-04-05)
Jens Blotevogel et al.
Environmental science & technology, 44(15), 5868-5874 (2010-07-09)
Models used to predict the fate of aqueous phase contaminants are often limited by their inability to address the widely varying redox conditions in natural and engineered systems. Here, we present a novel approach based on quantum chemical calculations that
Jens Blotevogel et al.
Environmental science & technology, 45(6), 2236-2242 (2011-02-22)
At contaminated field sites where active remediation measures are not feasible, monitored natural attenuation is sometimes the only alternative for surface water or groundwater decontamination. However, due to slow degradation rates of some contaminants under natural conditions, attenuation processes and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service