Skip to Content
Merck
All Photos(1)

Documents

52387

Sigma-Aldrich

Hexadecyltrimethylammonium hydroxide solution

~25% in methanol (T)

Synonym(s):

CTAOH solution, Cetrimonium hydroxide solution, Cetyltrimethylammonium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(OH)(CH3)3
CAS Number:
Molecular Weight:
301.55
Beilstein:
3723055
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

concentration

~25% in methanol (T)

refractive index

n20/D 1.372

SMILES string

[OH-].CCCCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C19H42N.H2O/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H2/q+1;/p-1

InChI key

WJLUBOLDZCQZEV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Hexadecyltrimethylammonium hydroxide solution is a commonly used phase transfer agent and surfactant solution employed in the synthesis of a variety of mesoporous materials, micelle complexes and transition-metal oxides.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A quantitative analysis of the effects of head group bulk on SN2 and E2 reactions in cationic micelles.
Brinchi L, et al.
Langmuir, 13(17), 4583-4587 (1997)
Salt-gel synthesis of porous transition-metal oxides.
Stein A, et al.
Chemistry of Materials, 7(2), 304-313 (1995)
Ordered mesoporous molecular sieves synthesized by a liquid-crystal template mechanism.
Kresge C T, et al.
Nature, 359(6397), 710-710 (1992)
Wormlike micelle assisted rod coating: A general method for facile fabrication of large-area conductive nanomaterial thin layer onto flexible plastics.
Xie J, et al.
ACS Applied Materials & Interfaces, 4(6), 2891-2896 (2012)
Studies on mesoporous materials: I. Synthesis and characterization of MCM-41.
Chen C Y, et al.
Microporous Mater., 2(1), 17-26 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service