Skip to Content
Merck
All Photos(3)

Key Documents

520861

Sigma-Aldrich

Potassium hexachloroplatinate(IV)

≥99.9% trace metals basis

Synonym(s):

Dipotassium hexachloroplatinate, Dipotassium platinum hexachloride, Platinic potassium chloride, Platinum potassium chloride, Potassium chloroplatinate, Potassium platinum(IV) chloride, Potassium platinum(IV) hexachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2PtCl6
CAS Number:
Molecular Weight:
485.99
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.9% trace metals basis

composition

Pt, 38-41%

reaction suitability

core: platinum
reagent type: catalyst

mp

250 °C (dec.) (lit.)

SMILES string

[K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.2K.Pt/h6*1H;;;/q;;;;;;2*+1;+4/p-6

InChI key

DPAIVKJGTXERIM-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

Potassium hexachloroplatinate(IV) can be used as a reactant to synthesize compounds such as chloropentaammineplatinum(IV), platinum triselenide, trans-dichloro(ethylenediamine-N,N′-di-3-propanoato)platinum(IV) and tetrachloro(O,O-dialkyl-ethylenediamine-N,N′-di-3-propanoato)platinum(IV) complexes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On the sulfides, selenides and tellurides of platinum.
Gr?nvold F, et al.
Acta Chemica Scandinavica, 14(9), 1879-1893 (1960)
Complex compounds of platinum (IV) and O, O-dialkyl-ethylenediamine-N, N?-di-3-propanoate ligands. A structural evidence for geometry of hydrolytic product of some esters.
Sabo T, et al.
Inorganic Chemistry Communications, 7(2), 241-244 (2004)
Outer-sphere one-equivalent changes in the vanadium (II) reductions of hexachloroplatinate (IV) and chloropentaammineplatinum (IV).
Bakac A, et al.
Inorganic Chemistry, 14(10), 2540-2543 (1975)
Hongliang Li et al.
Nature nanotechnology, 13(5), 411-417 (2018-03-21)
Exploring the interaction between two neighbouring monomers has great potential to significantly raise the performance and deepen the mechanistic understanding of heterogeneous catalysis. Herein, we demonstrate that the synergetic interaction between neighbouring Pt monomers on MoS2 greatly enhanced the CO2
Saurabh Samuchiwal et al.
Journal of hazardous materials, 402, 123835-123835 (2020-12-02)
A robust and efficient treatment process is required to address the problem of residual colour and avoid expensive post-treatment steps while dealing with textile effluents. In the present work, a novel microbial consortium enriched from textile effluent was used to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service