Skip to Content
Merck
All Photos(1)

Key Documents

205796

Sigma-Aldrich

Potassium tetrachloropalladate(II)

98%

Synonym(s):

Potassium palladium(II) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2PdCl4
CAS Number:
Molecular Weight:
326.43
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
98%
form:
crystals

Quality Level

Assay

98%

form

crystals

mp

105 °C (dec.) (lit.)

density

2.67 g/mL at 25 °C (lit.)

SMILES string

[K+].[K+].Cl[Pd--](Cl)(Cl)Cl

InChI

1S/4ClH.2K.Pd/h4*1H;;;/q;;;;2*+1;+2/p-4

InChI key

LGCKLDWLSVFMGL-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Potassium tetrachloropalladate(II) is a dark brown crystalline solid widely used as a Pd source in the field of catalysis, nanomaterial synthesis, and electronics.

Application

Potassium tetrachloropalladate(II) can be used:
  • As a precursor to synthesize Pd nanoparticles for catalytic degradation of organic pollutants and Pd-GO electrocatalyst in formic acid and ethanol oxidation.
  • To synthesize Pd–Pt alloy nanocrystals (NCs) with hollow structures by a galvanic replacement method with uniform Pd octahedral and cubic NCs as sacrificial templates. The hollow NCs exhibited higher ORR activities.
  • To synthesize immobilized Pd catalysts, a versatile method involves the layer-by-layer deposition of PAA and PEI-Pd(II) on alumina, followed by the reduction of Pd2+. This approach offers several benefits, including the stabilization of particles through the polyelectrolyte matrix, introduction of selectivity, and a significant reduction in undesired isomerization. Expanding the application of polyelectrolyte films holds promise for further enhancing selectivity in hydrogenation and other reactions.
  • To fabricate conductive and porous metal-organic frameworks(MOFs) for gas sensing applications and also to synthesize bimetallic Pd/SnO2 nanoparticles on metal organic framework (MOF) as an electrocatalyst for ethanol oxidation.
  • To prepare rigid macrocyclic pincer catalysts possessing polyaromatic ligands with enhanced catalytic activity.

Features and Benefits

Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar. Reacts with bis(dithiolates) to metal-bis(dithiolates) with applications in laser Q-switch materials, optical CD recording media, bar code material and superconductivity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L V Tat'ianenko et al.
Voprosy meditsinskoi khimii, 28(6), 126-131 (1982-11-01)
Mechanism of interaction between biologically active substances and membranes as well as and membrane-bound enzymes of two types: mitochondrial monoamine oxidase (MAO) from rat liver tissue and Ca2+, Mg+2-dependent ATPase from sarcoplasmic reticulum (SR) were studied. All the substances studied
Joris Muris et al.
Contact dermatitis, 67(2), 94-100 (2012-04-03)
Exposure to palladium (Pd) may lead to clinical allergic reactions. With frequent nickel (Ni) exposure and cross-reactivity between Ni and Pd at the T cell recognition level, positive Pd reactions on patch testing are surprisingly uncommon. PdCl(2) is often used
B L Iverson et al.
Nucleic acids research, 15(19), 7823-7830 (1987-10-12)
Reaction of DNA with K2PdCl4 at pH 2.0 followed by a piperidine workup produces specific cleavage at adenine (A) residues. Product analysis revealed the K2PdCl4 reaction involves selective depurination at adenine, affording an excision reaction analogous to the other chemical
Mathis, M. et al.
Chemistry of Materials, 10, 3568-3568 (1998)
Charles L Gaworski et al.
Inhalation toxicology, 20(2), 167-182 (2008-02-01)
The use of a palladium (Pd) catalyst has been proposed to promote combustion of tobacco, thereby reducing concentrations of certain toxic components of smoke, including polyaromatic hydrocarbons (PAHs). In the present work, toxicological comparisons were made using experimental cigarettes containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service