Skip to Content
Merck
All Photos(1)

Key Documents

50150

Sigma-Aldrich

Gly-Ala

≥99.0% (NT)

Synonym(s):

Glycyl-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CONHCH(CH3)COOH
CAS Number:
Molecular Weight:
146.14
Beilstein:
1724445
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (NT)

optical activity

[α]20/D −50±2°, c = 8% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

ign. residue

≤0.05% (as SO4)

mp

~230 °C (dec.)

application(s)

peptide synthesis

SMILES string

C[C@H](NC(=O)CN)C(O)=O

InChI

1S/C5H10N2O3/c1-3(5(9)10)7-4(8)2-6/h3H,2,6H2,1H3,(H,7,8)(H,9,10)/t3-/m0/s1

InChI key

VPZXBVLAVMBEQI-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Richmond Ayee et al.
Journal of Cancer, 11(7), 1737-1750 (2020-03-21)
Epstein Barr virus (EBV) is a cosmopolitan oncogenic virus, infecting about 90% of the world's population and it is associated to tumors originating from both epithelia and hematopoietic cells. Transmission of the virus is mainly through oral secretions; however, transmission
Fayez Kheir et al.
Cancers, 11(6) (2019-06-05)
Previous investigations proposed a link between the Epstein-Barr virus (EBV) and lung cancer (LC), but the results are highly controversial largely due to the insufficient sample size and the inherent limitation of the traditional viral screening methods such as PCR.
Xinyuan Wang et al.
Pharmacology & therapeutics, 211, 107526-107526 (2020-03-17)
The proteasome is a well-identified therapeutic target for cancer treatment. It acts as the main protein degradation system in the cell and degrades key mediators of cell growth, survival and function. The term "proteasome" embraces a whole family of distinct
Jiansen Du et al.
Biochemical and biophysical research communications, 490(3), 827-833 (2017-06-26)
Antibiotic resistance is becoming a global threat and overuse of antibiotics in aquaculture disease control worsens the situation. To reduce the risk of drug resistance developed in aquaculture, safer biocontrol programs are needed. Antivirulence therapy, with less chance for developing
P R Tulip et al.
The Journal of chemical physics, 131(1), 015103-015103 (2009-07-10)
We investigate the structure of the glycyl-l-alanine dipeptide in aqueous solution at a 1:20 peptide:water concentration via classical, atomistic molecular dynamics simulations using the CHARMM22 force field, and compare to recent neutron diffraction data [S. E. McLain, A. K. Soper

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service