Skip to Content
Merck
All Photos(1)

Key Documents

495328

Sigma-Aldrich

2-Chloro-5-methylpyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6ClN
CAS Number:
Molecular Weight:
127.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Quality Level

Assay

97%

refractive index

n20/D 1.53 (lit.)

bp

97 °C/30 mmHg (lit.)

density

1.169 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Cc1ccc(Cl)nc1

InChI

1S/C6H6ClN/c1-5-2-3-6(7)8-4-5/h2-4H,1H3

InChI key

VXLYOURCUVQYLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloro-5-methylpyridine is a pesticide intermediate. Its synthesis by many methods has been reported.

Application

2-Chloro-5-methylpyridine may be used in the synthesis of:
  • 2-methylthio-5-pyridinemethylene amine
  • 5-methyl-2,2′-bipyridine
  • 1-(5′-methyl-2,2′-bipyridin-5-yl)-2,5-dimethyl-1H-pyrrole

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and herbicidal activity of (Z)-ethoxyethyl 2-cyano-3-(2-methylthio-5-pyridylmethylamino) acrylates.
Wang QM, et al.
Heteroatom Chem., 15(1), 67-70 (2004)
Synthesis of Differently Disubstituted 2,2'-Bipyridines by a Modified Negishi Cross-Coupling Reaction.
Lutzen A, et al.
European Journal of Organic Chemistry, 2003(20), 3948-3957 (2003)
The preparation of 2-chloro-5-methyl-pyridine in airlift loop reactor.
Jianping W, et al.
Chemical Engineering Journal, 95(1), 33-36 (2003)
Synthesis of 5-Substituted 2,2'-Bipyridines from Substituted 2-Chloropyridines by a Modified Negishi Cross-Coupling Reaction.
Lutzen A and Hapke M.
European Journal of Organic Chemistry, 2002(14), 2292-2297 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service