Skip to Content
Merck
All Photos(1)

Documents

475750

Sigma-Aldrich

Bisphenol A propoxylate diglycidyl ether

PO/phenol 1

Synonym(s):

Bisphenol A bis(propylene glycol glycidyl ether )ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

composition

PO/phenol, 1

storage temp.

2-8°C

SMILES string

CC(C)(c1ccc(OCCCOCC2CO2)cc1)c3ccc(OCCCOCC4CO4)cc3

InChI

1S/C27H36O6/c1-27(2,21-5-9-23(10-6-21)30-15-3-13-28-17-25-19-32-25)22-7-11-24(12-8-22)31-16-4-14-29-18-26-20-33-26/h5-12,25-26H,3-4,13-20H2,1-2H3

InChI key

MROZYFGXESLRQQ-UHFFFAOYSA-N

General description

Bisphenol A propoxylate diglycidyl ether (BPDG) is an epoxy based resin that can be cured by ring opening mechanism of the oxirane groups in the monomer. It can be used to form structurally consistent and thermally stable composites for a variety of applications.

Application

BPDG can be used as a reinforcing material for silica aerogel for enhancing the thermo-mechanical properties of the composite. It can also be used to form hydrophilic epoxy networks which can potentially be used for surface coating and structural adhesives.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and properties of epoxy/polyaniline nanocomposites
Krakovsky I, et al.
Journal of Non-Crystalline Solids, 358(2), 414-419 (2012)
Polymer reinforced silica aerogels: effects of dimethyldiethoxysilane and bis (trimethoxysilylpropyl) amine as silane precursors
Randall JP, et al.
Journal of Material Chemistry A, 1(22), 6642-6652 (2013)
Structure and swelling behaviour of hydrophilic epoxy networks investigated by SANS
Krakovsky I, et al.
Polymer, 47(1), 218-226 (2006)
Very efficient surface relief holographic materials based on azobenzene-containing epoxy resins cured in films
Goldenberg LM, et al.
Journal of Materials Chemistry, 20(41), 9161-9171 (2010)
Enhanced thermal stability and structural characteristics of different MMT-Clay/epoxy-nanocomposite materials
Lakshmi MS, et al.
Polymer Degradation and Stability, 93(1), 201-213 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service