Skip to Content
Merck
All Photos(3)

Documents

909637

Sigma-Aldrich

NanoFabTx PLGA-nano

for synthesis of 100 and 200 nm particles

Synonym(s):

NanoFabTx, NanoFabTx reagent kit, PLGA nanoformulation kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
UNSPSC Code:
12161503
NACRES:
NC.25

description

PLGA nanoparticle screening kit for synthesis of 100 and 200 nm particles

Quality Level

application(s)

advanced drug delivery

storage temp.

2-8°C

General description

NanoFabTx PLGA-nano reagent kit is a nanoformulation kit designed for the synthesis of specifically sized, drug-encapsulating poly(lactic-co-glycolic acid) (PLGA) nanoparticles. The NanoFabTx PLGA-nano reagent kit provides reagents (PLGA and stabilizer) and protocols to synthesize polymeric nanoparticles 75 to 200 nm in size. One protocol describes how to use the kit with standard glassware using a nanoprecipitation or solvent-displacement method. The second protocol describes how to use the kit with the NanoFabTx Microfluidic - nano device kit (Cat.No. 911593) for the fabrication of nanoparticles using microfluidic methods.

Application

With NanoFabTx PLGA-nano reagent kit, users can simplify their methods for PLGA nanoparticle synthesis and their drug loading experiments. Our protocols describe how to tune the synthetic parameters to achieve monodisperse nanoparticles 75 nm in size, 200 nm in size, or any size in between. As a result, the kit enables users to synthesize and test different particle sizes and identify the ideal size and drug loading of PLGA nanocarriers for their research application. The resulting particles are biocompatible and biodegradable and can be further modified to target specific tissues or to ensure slow and sustained drug release. PLGA is a biocompatible and biodegradable polymer that is approved by the FDA for biomedical and pharmaceutical applications and that is used widely in drug delivery systems for sustained release of many different types of therapeutic molecules. PLGA-based nanocarriers can be used to encapsulate both hydrophobic drugs like curcumin and lipophilic drugs like docetaxel. PLGA nanocarriers synthesized with the NanoFabTx kits are suitable for biomedical research applications such as oncology, immuno-oncology, gene delivery, and vaccine delivery. Polymeric nanoparticles are useful for intravenous or systemic delivery of drug molecules. For example, polymeric nanocarriers improve drug dissolution and solubility of hydrophobic and lipophilic drugs. In addition, the small size of polymeric nanocarriers make them much more likely than larger microparticles to bypass the reticuloendothelial system and complement system, usually leading to longer circulation half-lives. In addition, nanoparticles can pass through the blood-brain barrier, through tumor vasculature, and can enter cells via pinocytosis.

Features and Benefits

  • Step-by-step protocols developed and tested by our formulation scientists
  • Flexible synthesis tool to create uniform and reproducible nanoparticles
  • Choose from standard glassware-based nanoprecipitation or microfluidic-based protocols
  • Optimized to make nanoparticles 75-200 nm nanoprecipitation or microfluidics with low polydispersity
  • Based on non-toxic, biodegradable polymers

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amit K Jain et al.
Critical reviews in therapeutic drug carrier systems, 28(1), 1-45 (2011-03-15)
Nanocarriers formulated with the US Food and Drug Administration-approved biocompatible and biodegradable polymer poly(lactic-co-glycolic acid) (PLGA) are being widely explored for the controlled delivery of therapeutic drugs, proteins, peptides, oligonucleotides, and genes. Surface functionalization of PLGA nanoparticles has paved the
Gaurav Kumar et al.
Critical reviews in therapeutic drug carrier systems, 29(2), 149-182 (2012-04-06)
Drug-loaded polylactide-co-glycolide (PLGA) nanoparticles have been extensively studied and have a practical impact on drug delivery. Many PLGA-based nanoformulations have reached different stages of preclinical development; however, they present distinct challenges for researchers. This review discusses some of the challenges
Anil Mahapatro et al.
Journal of nanobiotechnology, 9, 55-55 (2011-11-30)
Biodegradable nanoparticles (NPs) are gaining increased attention for their ability to serve as a viable carrier for site specific delivery of vaccines, genes, drugs and other biomolecules in the body. They offer enhanced biocompatibility, superior drug/vaccine encapsulation, and convenient release
Francesca Ungaro et al.
The Journal of pharmacy and pharmacology, 64(9), 1217-1235 (2012-08-14)
The aim of this review is to summarize the current state-of-the-art in poly(lactic-co-glycolic acid) (PLGA) carriers for inhalation. It presents the rational of use, the potential and the recent advances in developing PLGA microparticles and nanoparticles for pulmonary delivery. The
Fatemeh Sadat Tabatabaei Mirakabad et al.
Asian Pacific journal of cancer prevention : APJCP, 15(2), 517-535 (2014-02-27)
Poly (lactic-co-glycolic acid) (PLGA) is one of the most effective biodegradable polymeric nanoparticles (NPs). It has been approved by the US FDA to use in drug delivery systems due to controlled and sustained- release properties, low toxicity, and biocompatibility with

Articles

NanoFabTx™ platform accelerates drug development with ready-to-use formulations and microfluidic devices for particle synthesis.

NanoFabTx™ platform accelerates drug development with ready-to-use formulations and microfluidic devices for particle synthesis.

NanoFabTx™ platform accelerates drug development with ready-to-use formulations and microfluidic devices for particle synthesis.

NanoFabTx™ platform accelerates drug development with ready-to-use formulations and microfluidic devices for particle synthesis.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service