Skip to Content
Merck
All Photos(1)

Documents

475149

Sigma-Aldrich

3-(Trimethoxysilyl)propyl acrylate

92%, contains 100 ppm BHT as inhibitor

Synonym(s):

(3-Acryloyloxypropyl)trimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2(CH2)3Si(OCH3)3
CAS Number:
Molecular Weight:
234.32
Beilstein:
2046320
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

92%

contains

100 ppm BHT as inhibitor

refractive index

n20/D 1.429 (lit.)

bp

68 °C/0.4 mmHg (lit.)

density

1.055 g/mL at 25 °C (lit.)

SMILES string

CO[Si](CCCOC(=O)C=C)(OC)OC

InChI

1S/C9H18O5Si/c1-5-9(10)14-7-6-8-15(11-2,12-3)13-4/h5H,1,6-8H2,2-4H3

InChI key

KBQVDAIIQCXKPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(Trimethoxysilyl)propyl acrylate is a reactive monomer that is majorly used as a silane coupling agent for the formation of hybrid compounds. It facilitates the formation of a hydrophobic surface by lowering the surface tension and increasing the wettability.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

253.4 °F - closed cup

Flash Point(C)

123 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrophoretic concentration of proteins at laser-patterned nanoporous membranes in microchips
Song S, et al.
Analytical Chemistry, 76(15), 4589-4592 (2004)
Daichi Amano et al.
Dental materials journal, 29(1), 97-103 (2010-04-10)
The purpose of this study was to determine whether the brushing durability of a titanium dioxide coating on a polymethyl methacrylate (PMMA) substrate was improved by prior treatment with an acryloxypropyl trimethoxysilane-based agent. Titanium dioxide coatings were obtained by spray-coating
Reg Rehal et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 128, 43-53 (2018-11-25)
The mechanisms of membrane defence by lysylphosphatidylglycerol (LPG), were investigated using synthetic biomimetic mono- and bilayer models of methicillin resistant S. aureus ST239 TW, based on its lipid composition in both pH 7.4 (28% LPG) and pH 5.5 (51% LPG) cultures. These
Efrat Shukrun et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(8), 1800061-1800061 (2018-08-22)
Hybrid organic-inorganic sol gel inks that can undergo both condensation and radical polymerization are developed, enabling fabrication of complex objects by additive manufacturing technology, yielding 3D objects with superior properties. The 3D objects have very high silica content and are
Jukka Matinlinna et al.
Acta odontologica Scandinavica, 66(4), 250-255 (2008-07-16)
Four experimental blends of an organo-functional silane monomer with a non-functional cross-linking silane monomer (a novel silane system) were evaluated as adhesion promoters in an experiment in which a resin-composite cement was bonded to silica-coated titanium. 3-Acryloyloxypropyltrimethoxysilane (as constant 1.0

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service