Skip to Content
Merck
All Photos(1)

Key Documents

473820

Sigma-Aldrich

1,1,1-Trifluoro-3-iodopropane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICH2CH2CF3
CAS Number:
Molecular Weight:
223.96
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.42 (lit.)

bp

80 °C (lit.)

density

1.911 g/mL at 25 °C (lit.)

functional group

fluoro
iodo

SMILES string

FC(F)(F)CCI

InChI

1S/C3H4F3I/c4-3(5,6)1-2-7/h1-2H2

InChI key

ULIYQAUQKZDZOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,1,1-Trifluoro-3-iodopropane is a halogenated hydrocarbon. Its density at 25°C is 1.9110g/cm3.
Formation of polyfluoroalkylimidazolium salts.

Application

1,1,1-Trifluoro-3-iodopropane may be used in the preparation of quaternary salt, 1-methyl-3-trifluoropropylimidazolium iodide.
Used in a study of the quarternization of pyrazine, pyridazine, and pyrimidine.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis, 1072-1072 (2004)
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 27-27 (2015)
Synthesis, 1579-1579 (2003)
New dense fluoroalkyl-substituted imidazolium ionic liquids.
Singh RP, et al.
Tetrahedron Letters, 43(52), 9497-9499 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service