Skip to Content
Merck
All Photos(2)

Key Documents

402516

Sigma-Aldrich

1H-Pyrazole-1-carboxamidine hydrochloride

99%

Synonym(s):

1-Amidinopyrazole hydrochloride, Praxadine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4 · HCl
CAS Number:
Molecular Weight:
146.58
Beilstein:
5448758
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

167-170 °C (lit.)

SMILES string

Cl[H].NC(=N)n1cccn1

InChI

1S/C4H6N4.ClH/c5-4(6)8-3-1-2-7-8;/h1-3H,(H3,5,6);1H

InChI key

RBZRMBCLZMEYEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1H-Pyrazole-1-carboxamidine hydrochloride, a pyrazole derivative, is a heterocyclic compound. It is widely used in drug syntheses studies. Pyrazole ring forms the main core of various nonsteroidal anti-inflammatory drugs (NSAIDs) and antihypertensive drugs.

Application

1H-Pyrazole-1-carboxamidine hydrochloride may be used in the following studies:
  • Preparation of guanidylated hollow fiber membranes.
  • Guanylation of amines and in peptide synthesis.
  • Synthesis of bis-guanidinium-cholesterol derivatives.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hironori Izawa et al.
Biomolecules, 9(7) (2019-07-10)
In order to synthesize a promising material for developing a novel peptide/protein delivery system, guanidinylation of chitooligosaccharides with 1-amidinopyrazole hydrochloride was investigated herein. The production of guanidinylated chitooligosaccharides was demonstrated by infrared spectroscopy (IR), nuclear magnetic resonance (NMR), and elemental
J P Vigneron et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(18), 9682-9686 (1996-09-03)
Two cationic lipids, bis-guanidinium-spermidine-cholesterol (BGSC) and bis-guanidinium-trencholesterol (BGTC)-cholesterol derivatives bearing two guanidinium groups-have been synthesized and tested as artificial vectors for gene transfer. They combine the membrane compatible features of the cholesterol subunit and the favorable structural and high pKa
1H-Pyrazole-1-carboxamidine hydrochloride an attractive reagent for guanylation of amines and its application to peptide synthesis.
Bernatowicz MS, et al.
The Journal of Organic Chemistry, 57(8), 2497-2502 (1992)
Anna Yu Spivak et al.
Steroids, 154, 108530-108530 (2019-11-05)
A series of 34 new amine- and guanidine-functionalized derivatives of betulinic, ursolic, and oleanolic acids were synthesized and tested for their antimicrobial activity against the growth of four bacterial strains (Escherichia coli, Acinetobacter baumannii, Pseudomonas aeruginosa, and Staphylococcus aureus (MRSA))
Ekaterina K Ogurtsova et al.
Natural product communications, 10(7), 1171-1173 (2015-09-29)
The guanidine alkaloids, dihydropulchranin A (2), prepared from pulchranin A from the sponge Monanchora pulchra, and hexadecylguanidine (3), a synthetic analog of pulchranins, were studied for their TRPV channel-regulating activities. Compound 2 was active as an inhibitor of rTRPV1 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service