Skip to Content
Merck
All Photos(1)

Key Documents

469785

Sigma-Aldrich

Benzyl glycidyl ether

99%

Synonym(s):

(Benzyloxymethyl)oxirane, (±)-O-Benzylglycidol, 1-(Benzyloxy)-2,3-epoxypropane, 2-[(Phenylmethoxy)methyl]oxirane, 2-[[(Benzyloxy)methyl]oxy]oxirane, 3-(Benzyloxy)-1,2-epoxypropane, Glycidyl benzyl ether, [(Phenylmethoxy)methyl]oxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.5170 (lit.)

bp

70-73 °C/11 mmHg (lit.)

density

1.077 g/mL at 25 °C (lit.)

functional group

ether
phenyl

storage temp.

2-8°C

SMILES string

C(OCc1ccccc1)C2CO2

InChI

1S/C10H12O2/c1-2-4-9(5-3-1)6-11-7-10-8-12-10/h1-5,10H,6-8H2

InChI key

QNYBOILAKBSWFG-UHFFFAOYSA-N

General description

Benzyl glycidyl ether is a glycidyl derivative. Rac-/(R)-benzyl glycidyl ether participates in the synthesis of atactic and isotactic linear poly(benzyl 1,2-glycerol carbonate)s. Biocatalytic resolution of racemic benzyl glycidyl ether by Talaromyces flavus has been described. Resolution of benzyl glycidyl ether to (S)-benzyl glycidyl ether with 30% ee and (R)-3-benzyloxypropane-1,2-diol with 40% ee by whole Bacillus alcalophilus cells has been reported. It reacts with K-, K+(15-crown-5)2 to afford potassium glycidoxide and bibenzyl.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective cleavage of the linear ether bond in benzyl glycidyl ether and triphenylmethyl glycidyl ether by potassium alkalide as two-electron-transfer reagent.
Grobelny Z, et al.
Journal of Organometallic Chemistry, 660(1), 6-13 (2002)
Chun Wei et al.
Biotechnology letters, 34(8), 1499-1503 (2012-04-25)
Talaromyces flavus containing a constitutive epoxide hydrolase (EH) resolved racemic benzyl glycidyl ether and nine derivatives into their (R)-enantiomers. After optimization of the fermentation conditions, the specific EH activity and biomass concentration were improved from 13.5 U/g DCW and 14.8
Qingjie Zhang et al.
Nanomaterials (Basel, Switzerland), 8(9) (2018-09-12)
A surface-sizing technique was offered to take full advantage of multi-walled carbon nanotubes (MWCNTs) and epoxy resins. Two surface-sizing treated MWCNTs were obtained through a ball-milling treatment of amino-functionalized MWCNTs (MWCNT-NH₂) with n-butyl glycidylether (BuGE) and benzyl glycidylether (BeGE). These
Youlin Pan et al.
Chemistry, an Asian journal, 12(11), 1198-1203 (2017-03-21)
Cyclic azasilanes have been synthesized for the purpose of developing coupling agents appropriate for a variety of nanotechnologies including surface modification of nanoparticles, nanocrystals, mesoporous materials and substrates. N-Methyl-aza-2,2,4-trimethylsilacyclopentane is representative of this class of compounds. Preliminary data for the
Priya Saini et al.
World journal of microbiology & biotechnology, 33(5), 82-82 (2017-04-06)
In order to produce enantiomerically pure epoxides for the synthesis of value-added chemicals, a novel putative epoxide hydrolase (EH) sgeh was cloned and overexpressed in pET28a/Escherichia coli BL21(DE3). The 1047 bp sgeh gene was mined from Streptomyces griseus NBRC 13350 genome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service