Skip to Content
Merck
All Photos(1)

Key Documents

410829

Sigma-Aldrich

1,2-Epoxy-9-decene

96%

Synonym(s):

(7-Octenyl)oxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.442 (lit.)

density

0.842 g/mL at 25 °C (lit.)

functional group

allyl
ether

SMILES string

C=CCCCCCCC1CO1

InChI

1S/C10H18O/c1-2-3-4-5-6-7-8-10-9-11-10/h2,10H,1,3-9H2

InChI key

FCZHJHKCOZGQJZ-UHFFFAOYSA-N

General description

1,2-Epoxy-9-decene is an unsaturated epoxide. Hydrogenation of the C=C bond of 1,2-epoxy-9-decene over palladium nanoparticles, namely PdOAc,N has been reported.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Lu et al.
Chemistry of materials : a publication of the American Chemical Society, 23(18), 4227-4233 (2011-11-05)
Structurally diverse secondary amine-functionalized poly(propylene imine) (PPI) dendrimers capable of tunable nitric oxide (NO) release were synthesized in a straightforward, one-step manner using ring-opening or conjugate-addition reactions with propylene oxide (PO), styrene oxide (SO), acrylonitrile (ACN), poly(ethylene glycol) methyl ether
Reactivity versus Stability of Oxiranes under Palladium-Catalyzed Reductive Conditions.
Thiery E, et al.
European Journal of Organic Chemistry, 2009(7), 961-985 (2009)
Yuan Lu et al.
Biomacromolecules, 14(10), 3589-3598 (2013-08-22)
A series of amphiphilic nitric oxide (NO)-releasing poly(amidoamine) (PAMAM) dendrimers with different exterior functionalities were synthesized by a ring-opening reaction between primary amines on the dendrimer and propylene oxide (PO), 1,2-epoxy-9-decene (ED), or a ratio of the two, followed by
Alexandra Shakun et al.
Polymers, 11(7) (2019-07-03)
Detonation nanodiamonds, also known as ultradispersed diamonds, possess versatile chemically active surfaces, which can be adjusted to improve their interaction with elastomers. Such improvements can result in decreased dielectric and viscous losses of the composites without compromising other in-rubber properties
Claire Jeanquartier et al.
Langmuir : the ACS journal of surfaces and colloids, 24(24), 13957-13961 (2009-04-11)
A versatile two-step method has been developed that allows linking of biomolecules covalently to hydrogen-terminated group-IV semiconductors by means of epoxy-alkenes. First, the terminal C==C double bond of the alkene forms a covalent bond with the silicon, germanium, or diamond

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service