Skip to Content
Merck
All Photos(1)

Key Documents

469548

Sigma-Aldrich

1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione

98%

Synonym(s):

PTA, Pivaloyltrifluoroacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCOCH2COCF3
CAS Number:
Molecular Weight:
196.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

refractive index

n20/D 1.407 (lit.)

bp

138-141 °C (lit.)

density

1.129 g/mL at 25 °C (lit.)

functional group

fluoro
ketone

SMILES string

CC(C)(C)C(=O)CC(=O)C(F)(F)F

InChI

1S/C8H11F3O2/c1-7(2,3)5(12)4-6(13)8(9,10)11/h4H2,1-3H3

InChI key

BVPKYBMUQDZTJH-UHFFFAOYSA-N

Gene Information

mouse ... Mtap(66902)

General description

1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione (TFDMHD) is a 1,3-diketone.[1][2] It forms Er(3+) complexes. These complexes shows thermally activated slow relaxation of magnetization under a zero direct-current (DC) field.[3]

Application

1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione (TFDMHD) may be used in the synthesis of series of NNO ketoimines bearing trifluoromethyl substituents, via Schiff base condensation.[1] It may be used in the synthesis of octahedral Werner-type cobalt(II) complexes.[4] It was used in the preparation of D2-TFDMHD, by mixing with D2O.[2]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Baker et al.
Clinical rehabilitation, 31(1), 45-60 (2016-03-25)
To identify factors reported with negative and positive outcomes for caregivers of the traumatic brain injury and spinal cord injury cohorts, to investigate what interventions have been studied to support carers and to report what effectiveness has been found. Scoping
M Ramos Silva et al.
Dalton transactions (Cambridge, England : 2003), 44(3), 1264-1272 (2014-11-22)
The magnetic properties of two similar Er(3+) complexes have been investigated. [Er(tpm)3(bipy)] (Htpm = 1,1,1-trifluoro-5,5-dimethyl-2,4-hexanedione and bipy = 2,2'-bipyridine) displays thermally activated slow relaxation of magnetisation under a zero direct-current (DC) field. Under an applied HDC field of 1000 G
Conformation, molecular structure, and intramolecular hydrogen bonding of 1, 1, 1-trifluoro-5, 5-dimethyl-2, 4-hexanedione.
Vakili M, et al.
Journal of Molecular Structure, 1021, 102-111 (2012)
Anette Lohmander et al.
Journal of plastic surgery and hand surgery, 51(1), 27-37 (2017-02-22)
Adequate velopharyngeal function and speech are main goals in the treatment of cleft palate. The objective was to investigate if there were differences in velopharyngeal competency (VPC) and hypernasality at age 5 years in children with unilateral cleft lip and
Ippei Inoue et al.
Journal of bioscience and bioengineering, 122(5), 528-532 (2016-10-26)
We have developed an easy and rapid screening method of peptide aptamers with high affinity for a target material TiO2 using M13 phage-display and panning procedure. In a selection step, the phage-substrate complexes and Escherichia coli cells were directly applied

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service