Skip to Content
Merck
All Photos(1)

Documents

468312

Supelco

Poly(styrene-co-divinylbenzene) Polymeric Adsorbent

microspheres, 1400-2500 mesh

Synonym(s):

Polystyrene crosslinked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]x[CH2CH[C6H4(CHCH2)]]y
CAS Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
SB.52

product name

Poly(styrene-co-divinylbenzene), microspheres, 6.0-10.0 μm avg. part. size

form

microspheres

Quality Level

technique(s)

LPLC: suitable
thin layer chromatography (TLC): suitable

surface area

500 m2/g

matrix

Poly(styrene-co-divinylbenzene)

matrix active group

polymer

avg. part. size

6.0-10.0 μm

pore size

60 Å mean pore size

separation technique

reversed phase

SMILES string

C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.C=Cc3cccc(C=C)c3.C=Cc4ccccc4C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(styrene-co-divinylbenzene) are crosslinked microspheres composed of various concentrations of divinylbenzene from 5 to 75mol% based on styrene monomer. The gel-type PS beads are used as carrier to encapsulate metallocene catalysts.

Application

Poly(styrene-co-divinylbenzene) may be used:
  • in separation media for TLC, paper and LPL chromatography
  • as adsorbent
  • as matting agent for coatings, anti-blocking and lubrication additive for films and filler for ceramics, cosmetics and dental materials

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethylene (Co) Polymerization with Metallocene Catalysts Encapsulated in Gel-Type Poly (styrene-co-divinylbenzene) Beads.
Hong, Sung Chul, Ursula Rief, and Marc Oliver Kristen
Macromolecular Rapid Communications, 17, 1447-1454 (2001)
Fully crosslinked poly (styrene-co-divinylbenzene) microspheres by precipitation polymerization and their superior thermal properties.
Shim, Sang Eun, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 42, 835-845 (2004)
Sampling of airborne pesticides using Chromosorb 102.
T C Thomas et al.
Bulletin of environmental contamination and toxicology, 35(4), 460-465 (1985-10-01)
T C Thomas et al.
American Industrial Hygiene Association journal, 41(8), 599-601 (1980-08-01)
An organic polymeric solid, Chromosorb 102, has been reported to collect various pesticides from air. In this study the collection efficiency of Chromosorb 102 for chlordane was evaluated. Preparation of sampling tubes containing this medium for field sampling is also
The use of Chromosorb 102 to air samples pre-concentration in the determination of low concentrated dibutylphthalate.
L Wolska
Bulletin of the Institute of Maritime and Tropical Medicine in Gdynia, 39(1-2), 81-89 (1988-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service