Skip to Content
Merck
All Photos(1)

Key Documents

459607

Sigma-Aldrich

4,5-Dioctyloxy-1,2-benzenedicarbonitrile

97%

Synonym(s):

4,5-Dioctyloxyphthalonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)7O]2C6H2(CN)2
CAS Number:
Molecular Weight:
384.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

106-108 °C (lit.)

functional group

nitrile

SMILES string

CCCCCCCCOc1cc(C#N)c(cc1OCCCCCCCC)C#N

InChI

1S/C24H36N2O2/c1-3-5-7-9-11-13-15-27-23-17-21(19-25)22(20-26)18-24(23)28-16-14-12-10-8-6-4-2/h17-18H,3-16H2,1-2H3

InChI key

SSVVHLRGCJLWCP-UHFFFAOYSA-N

Related Categories

Application

4,5-Dioctyloxy-1,2-benzenedicarbonitrile (4,5-Dioctyloxyphthalonitrile) may be used for the preparation of new unsymmetrically substituted DB24C8-phthalocyanines.[1] It may be used for the synthesis of tetrathiafluvalene (TTF)-annulated symmetrical and unsymmetrical phthalocyanines.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and Optical and Electrochemical Properties of Phthalocyanines with the Ttf Unit.
Kimura T, et al.
Phosph. Sulfur Relat. Elem., 188(4), 408-412 (2013)
Martinez-Diaz et al.
Organic letters, 2(8), 1057-1060 (2000-05-11)
[formula: see text] New unsymmetrically substituted DB24C8-phthalocyanines, which are able to form complexes with suitable dialkylammonium cations, have been prepared. These complexes most probably have a pseudorotaxane geometry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service