All Photos(3)
About This Item
Linear Formula:
O2NC6H3-1,2-(CN)2
CAS Number:
Molecular Weight:
173.13
Beilstein:
1877554
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Recommended Products
Assay
99%
form
powder, crystals or chunks
mp
142-144 °C (lit.)
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
[O-][N+](=O)c1ccc(C#N)c(c1)C#N
InChI
1S/C8H3N3O2/c9-4-6-1-2-8(11(12)13)3-7(6)5-10/h1-3H
InChI key
NTZMSBAAHBICLE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Shambhulinga Aralekallu et al.
Dalton transactions (Cambridge, England : 2003), 49(42), 15061-15071 (2020-10-27)
An indirect electrochemical detoxification and detection platform has been demonstrated for toxic hexavalent chromium (Cr(vi)) based on the biologically important N-4 macrocycle. The research work describes a simple, green, low-cost and potential way for the synthesis of a new N-4
Eradication of solid tumors by chemodynamic theranostics with H2O2-catalyzed hydroxyl radical burst.
Nana Wang et al.
Theranostics, 11(5), 2334-2348 (2021-01-28)
Activatable theranostics, integrating high diagnostic accuracy and significant therapeutic effect, holds great potential for personalized cancer treatments; however, their chemodynamic modality is rarely exploited. Herein, we report a new in situ activatable chemodynamic theranostics PAsc/Fe@Cy7QB to specifically recognize and eradicate
Ruiqi Yang et al.
Scientific reports, 6, 36434-36434 (2016-11-09)
Dielectric film with ultrahigh thermal stability based on crosslinked polyarylene ether nitrile is prepared and characterized. The film is obtained by solution-casting of polyarylene ether nitrile terminated phthalonitrile (PEN-Ph) combined with post self-crosslinking at high temperature. The film shows a
Wael M Darwish et al.
Journal of photochemistry and photobiology. B, Biology, 203, 111777-111777 (2020-01-14)
Despite the high in vitro efficacy of photodynamic therapeutics, lack of tumor targeting significantly reduces their in vivo efficacy and thus limits their clinical use. Photoimmunotherapy (PIT) is a new synthetic strategy to target and treat cancer by photodynamic therapy
Wenjuan Zhang et al.
Analytical and bioanalytical chemistry, 410(25), 6653-6661 (2018-07-27)
In this work, poly(glycidyl methacrylate-ethyleneglycol dimethacrylate) monolith functionalized with copper tetra(N-carbonylacrylic) aminephthalocyanine and iminodiacetic acid was successfully synthesized. Owing to hydrogen bonding and hydrophilic interactions, the monolith exhibited good performance for glycopeptide enrichment. When the tryptic digests of horseradish peroxidase
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service