Skip to Content
Merck
All Photos(1)

Key Documents

450820

Sigma-Aldrich

Copper(II) sulfide

powder, 99.99% trace metals basis

Synonym(s):

Copper Blue, Copper monosulfide, Cupric sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuS
CAS Number:
Molecular Weight:
95.61
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder

mp

220 °C (dec.) (lit.)

density

4.6 g/mL at 25 °C (lit.)

SMILES string

S=[Cu]

InChI

1S/Cu.S

InChI key

BWFPGXWASODCHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cu63 NMR, X ray photoelectron spectroscopy,2 copper sulfide CuS was studied in detail.

Packaging

Packaged in ampules

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caifeng Ding et al.
Biosensors & bioelectronics, 25(5), 1082-1087 (2009-10-27)
An ultrasensitive chemiluminescence method based on the Au nanoparticles amplification for the quantitative detection of single-nucleotide polymorphisms (SNPs) in genomic DNA was accomplished by the DNA polymerase I (Klenow fragment)-induced coupling of the nucleotide-modified nanoparticle probe to the mutant sites
Nirupam Banerjee et al.
Dalton transactions (Cambridge, England : 2003), 39(41), 9789-9793 (2010-09-11)
Well uniform microspheres of phase pure Covellite were synthesized through a simple hydrothermal approach using poly vinyl pyrrolidone (PVP) as surfactant. The micro-spheres were constituted of numerous self-organized knitted nano-ribbons of ∼ 30 nm thickness. The effect of conc. PVP
Mrinmoyee Basu et al.
Environmental science & technology, 44(16), 6313-6318 (2010-08-14)
Blue solution of copper(II) acetylacetonate complex, [Cu(acac)(2)] in dichloromethane (DCM) and an aqueous alkaline solution of thioacetamide (TAA) constitute a biphasic system. The system in a screw cap test tube under a modified hydrothermal (MHT) reaction condition produces a greenish
Hydrophilic flower-like CuS superstructures as an efficient 980 nm laser-driven photothermal agent for ablation of cancer cells.
Qiwei Tian et al.
Advanced materials (Deerfield Beach, Fla.), 23(31), 3542-3547 (2011-07-08)
K Pedersen
Journal of applied microbiology, 108(3), 1094-1104 (2009-12-18)
To investigate the relationships between sulfate-reducing bacteria (SRB), growth conditions, bentonite densities and copper sulfide generation under circumstances relevant to underground, high-level radioactive waste repositories. Experiments took place 450 m underground, connected under in situ pressure to groundwater containing SRB.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service