Skip to Content
Merck
All Photos(1)

Documents

566284

Sigma-Aldrich

Copper(I) oxide

≥99.99% trace metals basis, anhydrous

Synonym(s):

Cuprous oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu2O
CAS Number:
Molecular Weight:
143.09
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

Assay

≥99.99% trace metals basis

form

powder

density

6 g/mL at 25 °C (lit.)

SMILES string

[Cu]O[Cu]

InChI

1S/2Cu.O

InChI key

BERDEBHAJNAUOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Copper(I) oxide is a weak inorganic base that can be used to activate halides for nucleophilic substitution reaction. It is also used in decarboxylation and cyclo condensation reactions.

Application

Copper(I) oxide can be used:
  • As a precursor to prepare nanoparticles, nanocrystals, and quantum dots for various applications.
  • To catalyze the N-arylation of azoles under aqueous conditions using phase-transfer catalyst.
  • To prepare reduced graphene oxide-based nano copper composites applicable as a sensor for the detection of dopamine.
  • As a copper precursor to prepare Cu2O-TiO2 nanoparticle-based composite materials with aluminosilicate geopolymers applicable in the removal of organic pollutants from water.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper(I) oxide
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2009)
Investigation of copper (I) oxide quantum dots by near-edge X-ray absorption fine structure spectroscopy
Nachimuthu P, et al.
Chemistry of Materials, 15(20), 3939-3946 (2003)
Synthesis and properties of novel photoactive composites of P25 titanium dioxide and copper (I) oxide with inorganic polymers
Falah M and MacKenzie KJD
Ceramics International, 41(10), 13702-13708 (2015)
A ligand-free copper (I) oxide catalyzed strategy for the N-arylation of azoles in water
Yong FF, et al.
Tetrahedron Letters, 52(11), 1161-1164 (2011)
Copper (I) oxide nanocrystals-one step synthesis, characterization, formation mechanism, and photocatalytic properties
Singhal A, et al.
European Journal of Inorganic Chemistry, 2013(14), 2640-2651 (2013)

Articles

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service