Skip to Content
Merck
All Photos(3)

Key Documents

44890

Sigma-Aldrich

Iron

puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)

Synonym(s):

Fe

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

puriss. p.a.

Assay

≥99.5% (RT)

quality

low in magnesium and manganese compounds

reaction suitability

core: iron
reagent type: catalyst

resistivity

9.71 μΩ-cm

impurities

≤0.01% total nitrogen (N)

grain size

5-9 μm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

cation traces

As: ≤2 mg/kg
Ca: ≤50 mg/kg
Cd: ≤10 mg/kg
Co: ≤50 mg/kg
Cr: ≤10 mg/kg
Cu: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤10 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Application

Iron (carbonyl) powder has been used:
  • In the preparation of copper on iron (Cu/Fe) bimetallic catalytic system to synthesize β-aminoenones and 3,5-disubstituted pyrazoles.[1]
  • As the main component of magnetorheological fluids.[2][3]
  • To produce Fe-B-C composites from boron carbide using plasma sintering.[4]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Self-heat. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Effect of particle size on shear stress of magnetorheological fluids.
Sarkar, Chiranjit and Hirani, Harish
Smart Science, 3(2), 65-73 (2015)
The role of surface tension in magnetorheological adhesion.
Orellana, Carlos S and Jaeger, Heinrich M
Soft Matter, 9(35), 8519-8525 (2013)
Fe-B-C composites produced using spark plasma sintering.
Rokebrand, P and Sigalas, I
International Journal of Refractory Metals & Hard Materials, 49(35), 320-326 (2015)
Copper on iron promoted one-pot synthesis of β-aminoenones and 3, 5-disubstituted pyrazoles.
Kovacs, Szabolcs and Novak, Zoltan
Tetrahedron, 69(43), 8987-8993 (2013)
Jia-Wei Yang et al.
Frontiers in cell and developmental biology, 7, 275-275 (2019-12-04)
Current research has enabled the use of microphysiological systems and creation of models for alveolar and pulmonary diseases. However, bottlenecks remain in terms of medium- and long-term regulation of cell cultures and their functions in microchannel systems, as well as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service