Skip to Content
Merck
All Photos(3)

Key Documents

209309

Sigma-Aldrich

Iron

powder, −325 mesh, 97%

Synonym(s):

Fe

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

powder

reaction suitability

core: iron
reagent type: catalyst

resistivity

9.71 μΩ-cm

particle size

−325 mesh

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Application

Iron is generally used as a catalyst for wide variety of organic transformations. It has been used for the synthesis of magnetic polydimethylsiloxane (PDMS) for automated microcontact printing of biomolecules.[1]
Iron can also be used:
  • For the removal of arsenate from water.[2]
  • To synthesize Fe(III) iodide catalyst for thioglycosylation of peracetylated saccharides.[3]
  • As a catalyst for the graphitization for microscale radiocarbon analysis of small samples.[4]
  • For the removal of nitroaromatic pesticides by reduction.[5]

Physical form

Hydrogen reduced

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Self-heat. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 6

1 of 6

Arsenate removal by zero valent iron: batch and column tests.
Biterna M, et al.
Journal of Hazardous Materials, 149(3), 548-552 (2007)
Fabrication of biomolecule microarrays for cell immobilization using automated microcontact printing.
Cell-Based Microarrays, 83-95 (2018)
Diastereoselective thioglycosylation of peracetylated glycosides catalyzed by in situ generated iron (III) iodide from elemental iodine and iron.
Weng SS
Tetrahedron Letters, 50(46), 6414-6417 (2009)
Technical progress in AMS microscale radiocarbon analysis.
Uchida M, et al.
Nucl. Instrum. Methods Phys. Res. Sect. B, 223(46), 313-317 (2004)
Reduction of nitroaromatic pesticides with zero-valent iron.
Keum Y S and Li Q X
Chemosphere, 54(3), 255-263 (2004)

Articles

Mechanical alloying is a “brute force” method of affecting alloying and chemical reactions. The mixture of reactant powders and several balls are placed in the milling jar of a high-energy ball mill, for example, a shaker mill or a planetary mill.

Questions

  1. Are the particles spherical or some other morphology?

    1 answer
    1. The material is supplied with a crushed irregular shape. Any other morphology information is not determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service