Skip to Content
Merck
All Photos(3)

Key Documents

435325

Sigma-Aldrich

Poly(acrylic acid) partial potassium salt

<1000 μm particle size

Synonym(s):

Poly(acrylic acid) potassium salt, Potassium polyacrylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

impurities

<600 ppm residual acrylic acid

particle size

<1000 μm

density

0.4 g/mL at 25 °C (lit.)

SMILES string

[K+].OC(=O)C=C.[O-]C(=O)C=C

InChI

1S/C3H4O2.K/c1-2-3(4)5;/h2H,1H2,(H,4,5);/q;+1/p-1

InChI key

ZUBIJGNKOJGGCI-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of chitosan-g-poly (acrylic acid)/attapulgite superabsorbent composites
Zhang J, et al.
Carbohydrate Polymers, 68(2), 367-374 (2007)
Studies on poly (acrylic acid)/attapulgite superabsorbent composite. I. Synthesis and characterization
Li An, et al.
Journal of Applied Polymer Science, 92(3), 1596-1603 (2004)
Superabsorbent Polymer Materials: A Review
Zohuriaan-Mehr MJ and Kabiri K
Iranian Polymer Journal, 17(6), 451-477 (2008)
Study on Na and K salts of PAA as corrosion inhibitors
Mishra S, et al.
J. Sci. Ind. Res., 64, 684-687 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service