430072
Poly(styrene-co-butadiene)
butadiene 4 wt. %, melt index 6 g/10 min (200°C/5kg)
About This Item
Recommended Products
form
pellets
Quality Level
melt index
6 g/10 min (200°C/5kg)
composition
butadiene, 4 wt. %
hardness
105 (Rockwell R, ASTM D 785)
transition temp
softening point 103 °C (Vicat, ASTM D 1525)
density
1.04 g/mL at 25 °C
SMILES string
C=CC=C.C=Cc1ccccc1
InChI
1S/C8H8.C4H6/c1-2-8-6-4-3-5-7-8;1-3-4-2/h2-7H,1H2;3-4H,1-2H2
InChI key
MTAZNLWOLGHBHU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Features and Benefits
Physical form
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service