All Photos(2)
About This Item
Linear Formula:
(CH2CH=CHCH2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
mol wt
average Mn ~5,000
Quality Level
composition
cis- and trans-1,4, 80%
vinyl, 20%
density
0.9 g/mL at 25 °C (lit.)
SMILES string
C\C=C\C
InChI
1S/C4H8/c1-3-4-2/h3-4H,1-2H3/b4-3+
InChI key
IAQRGUVFOMOMEM-ONEGZZNKSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Influence of Biogenic Material Content on the Biodegradability of Styrene-Butadiene Composites with Incorporated Chlorella vulgaris Biomass.: This study explores the impact of biogenic materials on the biodegradability of styrene-butadiene composites. It highlights the potential of polybutadiene composites in developing environmentally friendly materials, particularly in the context of high resilience rubber applications (Bumbac et al., 2024).
- Exploring the synergistic effect of carboxymethyl cellulose and chitosan in enhancing thermal stability of polyurethanes through statistical mixture design approach.: Although focused on polyurethanes, this research provides insights into the thermal stability enhancement techniques that could be applied to polybutadiene in medical device materials and life science manufacturing polymers (Javaid et al., 2024).
- Self-Amplified HF Release and Polymer Deconstruction Cascades Triggered by Mechanical Force.: This article discusses the deconstruction of polymers triggered by mechanical force, offering valuable knowledge for developing synthetic rubber properties for healthcare applications, especially in polybutadiene-based medical equipment (Hu et al., 2024).
- Rheological and Mechanical Characterization of 3D-Printable Solid Propellant Slurry.: The research covers the mechanical properties and 3D-printing capabilities of polymer slurries, pertinent to the development of high resilience polybutadiene for life science manufacturing (Zumbo et al., 2024).
- Rubber Oxygenase Degradation Assay by UV-Labeling and Gel Permeation Chromatography.: This study provides insights into the degradation mechanisms of rubber materials, relevant for improving the durability and properties of polybutadiene rubber in medical device innovation (Adjedje et al., 2024).
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Zhi-ping Huang et al.
Journal of hazardous materials, 229-230, 251-257 (2012-06-26)
Migration appeared in the interfaces of nitrate ester plasticized polyether (NEPE) based propellant/hydroxyl-terminated polybutadiene (HTPB) based liner/ethylene propylene terpolymer (EPDM) based insulation was studied by aging at different temperatures. The migration components were extracted with solvent and determined by high
Rachel Calvet et al.
Journal of chromatography. A, 1253, 164-170 (2012-07-24)
In this study, the coupling of IGC-ID analysis and carbon blacks impregnation with increasing ratios of polymer allow us to highlight the affinity of the polymer to the solid surface. The variations of the dispersive component of the surface energy
Bounphanh Tonpheng et al.
Physical chemistry chemical physics : PCCP, 13(33), 15047-15054 (2011-07-22)
The thermal conductivity κ, heat capacity per unit volume ρc(p) and glass transition behaviour under pressure have been established for medium and high vinyl content polybutadiene PB with molecular weights 2600 and 100,000 and their highly cross-linked (ebonite) states obtained
Gregory P Robbins et al.
Langmuir : the ACS journal of surfaces and colloids, 26(17), 14089-14096 (2010-08-14)
The polymersome, a fully synthetic cell mimetic, is a tunable platform for drug delivery vehicles to detect and treat disease (theranostics). Here, we design a leuko-polymersome, a polymersome with the adhesive properties of leukocytes, which can effectively bind to inflammatory
Joan M Widin et al.
Journal of the American Chemical Society, 134(8), 3834-3844 (2012-01-28)
Controlled/"living" polymerizations and tandem polymerization methodologies offer enticing opportunities to enchain a wide variety of monomers into new, functional block copolymer materials with unusual physical properties. However, the use of these synthetic methods often introduces nontrivial molecular weight polydispersities, a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service