Skip to Content
Merck
All Photos(2)

Key Documents

422967

Sigma-Aldrich

4-Aminothiophenol

≥97.0%

Synonym(s):

4-Aminobenzenethiol, 4-Aminophenyl mercaptan, 4-Mercaptoaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4SH
CAS Number:
Molecular Weight:
125.19
Beilstein:
906904
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

bp

140-145 °C/16 mmHg (lit.)

mp

37-42 °C (lit.)

storage temp.

2-8°C

SMILES string

Nc1ccc(S)cc1

InChI

1S/C6H7NS/c7-5-1-3-6(8)4-2-5/h1-4,8H,7H2

InChI key

WCDSVWRUXWCYFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Aminothiophenol is a bifunctional aromatic thiol used to form self-assembled monolayers for biosensors.

4-Aminothiophenol (p-aminothiophenol, PATP) is an aromatic thiol. The self-assembly of silver and gold nano particles which are interconnected with 4-ATP to form metal-molecule-metal sandwich architecture has been characterized by surface enhanced Raman scattering (SERS) using an off-surface plasmon resonance condition. Chemical transformation of PATP to 4,4-dimercaptoazobenzene (DMAB) has been extensively studied.

Application

4-Aminothiophenol (4-ATP) was employed as bifunctional molecule to functionalize gold nanoparticles (NPs) used in surface-enhanced Raman spectroscopy (SERS) detection of thrombin by protein recognition.

It may be used in the following studies:
  • As a water insoluble ligand in the synthesis of pectin–4-aminothiophenol conjugates.
  • To functionalize gold nanorods (GNR) in the development of GNR nitrite sensor.
  • To form self-assembled monolayers (SAMs) of 4-aminothiophenol in the development of DNA biosensor.
  • As a linker to thiol (SH) functionalize multiwalled carbon nanotubes (MWCNTs).
  • To generate amino terminated self-assembly monolayers(SAMs) on platinum electrode which forms the base for attachment of single-wall carbon nano tubes (SWCTs).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface-enhanced Raman scattering of 4-aminothiophenol self-assembled monolayers in sandwich structure with nanoparticle shape dependence: off-surface plasmon resonance condition.
Hu X, et al.
The Journal of Physical Chemistry C, 111(19), 6962-6969 (2007)
SERS detection of thrombin by protein recognition using functionalized gold nanoparticles.
Bizzarri AR, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 3(4), 306-310 (2007)
4-Aminothiophenol Self-Assembled Monolayer for the Development of a DNA Biosensor Aiming the Detection of Cylindrospermopsin Producing Cyanobacteria.
Valerio E, et al.
Electroanalysis, 20(22), 2467-2474 (2008)
Application of coulometric FFT cyclic voltammetry for determination of cholesterol based on cholesterol oxidase nano-biosensor.
Norouzi P.
Analytical and Bioanalytical Electrochemistry, 4(1), 70-82 (2010)
Coral Salvo-Comino et al.
Materials (Basel, Switzerland), 13(3) (2020-02-09)
Phenolic compounds such as catechol are present in a wide variety of foods and beverages; they are of great importance due to their antioxidant properties. This research presents the development of a sensitive and biocompatible molecular imprinted sensor for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service