Skip to Content
Merck
All Photos(3)

Key Documents

419702

Sigma-Aldrich

2-Mercaptopyridine-3-carboxylic acid

technical grade

Synonym(s):

2-Mercaptonicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2S
CAS Number:
Molecular Weight:
155.17
Beilstein:
119029
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

form

powder

mp

263-265 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1cccnc1S

InChI

1S/C6H5NO2S/c8-6(9)4-2-1-3-7-5(4)10/h1-3H,(H,7,10)(H,8,9)

InChI key

WYKHFQKONWMWQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Mercaptopyridine-3-carboxylic acid may be used for the synthesis of poly[[diaquabis([μ]2-4,4 ′-bipyridyl)iron(II)] bis{2-[(3-carboxypyridin-2-yl)disulfanyl]nicotinate}] and 2-(2-carboxyethylthio)nicotinic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Partenhauser et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 105(3), 551-559 (2015-11-28)
Assessment of preactivated and thiolated silicone oils as potential long-term vitreous replacement. Thioglycolic acid (TGA) and 3-mercaptopropionic acid (MPA) were covalently coupled to amino-modified silicone oil and subsequently preactivated with 2-mercaptonicotinic acid (2-MNA). Each silicone thiomer was evaluated in view
Ioannis I Verginadis et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 42(3), 253-261 (2010-12-07)
Nowadays, investigation for possible therapeutic applications of various metal-based drugs attracts the scientific interest worldwide. The triorganotin compound bis[triphenyltin(IV)](3-carboxy-pyridine-2-thionato) (SnMNA), was tested for its anti-proliferative and antitumor activities. Cytotoxic activity was assessed by Trypan blue and 3-(4.5-dimethylthiazol-2-yl)-2.5-diphenyltetrazolium bromide assay (MTT).
Xiao-Juan Wang et al.
Acta crystallographica. Section E, Structure reports online, 66(Pt 6), o1298-o1298 (2010-01-01)
The title compound, C(8)H(7)NO(4)S·H(2)O, was obtained by reaction of 2-mercaptopyridine-3-carboxylic acid with chloro-acetic acid. In the mol-ecular structure, the dihedral angle between the two least-squares planes defined by the pyridine ring and the carb-oxy group is 8.32 (9)°. The carboxy-methyl-sulfanyl group
Aamir Jalil et al.
Molecular pharmaceutics, 15(8), 3527-3534 (2018-07-27)
The aim of this study was to synthesize iodine containing polymeric excipients for mucosal treatment of microbial infection exhibiting a prolonged mucosal residence time by forming an adhesive gel on the mucosal surface. In order to achieve this aim, 2-(2
Maurizio Casarin et al.
Inorganic chemistry, 49(9), 4099-4108 (2010-04-02)
A Zn(II) 2-mercaptonicotinate coordination polymer (Zn1), with Zn(II) ions chelated by both sulfur and oxygen in a distorted square pyramidal environment, and a molecular Zn(II) 2-hydroxynicotinate complex (Zn2) were synthesized by the reaction of zinc acetylacetonate with 2-mercaptonicotinic (Zn1) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service