Skip to Content
Merck
All Photos(3)

Key Documents

419281

Sigma-Aldrich

Sodium carboxymethyl cellulose

average Mw ~250,000, degree of substitution 1.2

Synonym(s):

Carboxymethylcellulose sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

powder

autoignition temp.

698 °F

mol wt

average Mw ~250,000

extent of labeling

1.2 carboxymethyl groups per anhydroglucose unit

mp

270 °C (dec.)

SMILES string

[Na].OC(C(O)C(O)C=O)C(O)CO.OC(=O)C

InChI

1S/C6H12O6.C2H4O2.Na/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4;/h1,3-6,8-12H,2H2;1H3,(H,3,4);

InChI key

DPXJVFZANSGRMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium carboxymethyl cellulose (CMC) belongs to the class of anionic linear structured cellulose. Its components consist of polysaccharide composed of fibrous tissues of plants. It is a water soluble polymer which can be used as a polyelectrolyte cellulose derivative.

Application

CMC can be used for a wide range of application such as:
  • food processing, textiles, detergents, and drugs
  • a stabilizer in the preparation of nanoparticles
  • a binder for the formation of electrodes

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela M Correia et al.
Materials (Basel, Switzerland), 13(10) (2020-05-21)
This study reports a versatile method for the development of cellulose nanocrystals (CNCs) and water-soluble cellulose derivatives (methyl cellulose (MC), hydroxypropyl cellulose (HPC), and sodium carboxymethyl cellulose (NaCMC)) films comprising the ionic liquid (IL) 2-hydroxy-ethyl-trimethylammonium dihydrogen phosphate ([Ch][DHP]) for actuator
Sodium carboxymethyl cellulose a potential binder for Si negative electrodes for Li-ion batteries
Li J, et al.
Electrochemical and Solid-State Letters, 10(2), A17-A20 (2007)
Silver nanoparticles as pigments for water-based ink-jet inks
Magdassi S, et al.
Chemistry of Materials, 15(11), 2208-2217 (2003)
In situ synthesis of polyaniline/sodium carboxymethyl cellulose nanorods for high-performance redox supercapacitors
Peng H, et al.
Journal of Power Sources, 211, 40-45 (2012)
Stabilization of Fe- Pd nanoparticles with sodium carboxymethyl cellulose for enhanced transport and dechlorination of trichloroethylene in soil and groundwater
He F, et al.
Industrial & Engineering Chemistry Research, 46(1), 29-34 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service