Skip to Content
Merck
All Photos(3)

Documents

C5013

Sigma-Aldrich

Carboxymethylcellulose sodium salt

High viscosity

Synonym(s):

Sodium carboxymethylcellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

wood (pulp)

form

powder

color

light yellow

useful pH range

(20 °C, 10 g/L, 68 °F neutral)

viscosity

1500-3000 cP, 1 % in H2O(25 °C)(lit.)

mp

> 300.05  °C ((> 572.09 °F ))

solubility

H2O: 10 mg/mL

cation traces

Na: 6.5-9.5%

storage temp.

room temp

InChI

1S/C6H12O6.C2H4O2.Na/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4;/h1,3-6,8-12H,2H2;1H3,(H,3,4);

InChI key

DPXJVFZANSGRMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is a high viscosity carboxymethylcellulose (CMC); the viscosity of a 1% solution in water at 25 oC is 1500-3000 centipoise (cps). The viscosity is both concentration and temperature dependent. As the temperaure increases, the viscosity decreases. As the concentration increases, the viscosity increases.Low, medium and high viscosity CMCs are all used as suspending agents. Low viscosity CMC is usually used in ""thin"" aqueous solutions. Medium viscosity CMC is used to make solutions that look like a syrup. High viscosity CMC is used to make a mixture, which resembles a cream or lotion.

Application

Soluble carboxymethylcelluloses (CM-cellulose; CMC) available in varying viscosities are used as viscosity modifiers (thickeners) to stabilize emulsions and as a chemical dispersants of oils and other carbon structures such as nanotubes. CMCs are used in the development of biostructures such as biofilms, emulsions and nanoparticles for drug delivery. Carboxymethylcellulose, high viscosity, may be used to make solutions the consistency of creams or lotions.

Preparation Note

The product is soluble in water (10 mg/ml). The key to dissolving carboxymethylcellulose is to add the solid carefully to the water so that it is well dispersed (well-wetted). Adding the solid in portions may be necessary. Adding water to the dry solid produces a "clump" of solid that is very difficult to dissolve; the solid must be added to the water. Stir gently or shake intermittently; do not stir constantly with a magnetic stirring bar. High heat is not needed and may actually slow down the solubilization process. A mixing device, such as an impeller-type agitator which produces a vortex, would allow the powder to be drawn into the liquid, but it may produce some shearing. Note that this high viscosity CMC is usually used at concentrations of less than 20 mg/ml, but it is soluble in water at up to 50 mg/ml; heat is required to prepare a 50 mg/ml solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shannah K Witchey et al.
Neurotoxicology, 74, 139-148 (2019-06-30)
Bisphenol A (BPA) is a well-characterized endocrine disrupting chemical (EDC) used in plastics, epoxy resins and other products. Neurodevelopmental effects of BPA exposure are a major concern with multiple rodent and human studies showing that early life BPA exposure may
Sarah A Johnson et al.
Hormones and behavior, 80, 139-148 (2015-10-06)
Bisphenol A (BPA) is a ubiquitous industrial chemical used in the production of a wide variety of items. Previous studies suggest BPA exposure may result in neuro-disruptive effects; however, data are inconsistent across animal and human studies. As part of
Shafiqul I Chowdhury et al.
Vaccines, 9(1) (2021-01-17)
The bovine respiratory disease complex (BRDC) remains a major problem for both beef and dairy cattle industries worldwide. BRDC frequently involves an initial viral respiratory infection resulting in immunosuppression, which creates a favorable condition for fatal secondary bacterial infection. Current
Wai-Hong Wu et al.
Virology journal, 12, 140-140 (2015-09-13)
Infection by any one of 15 high risk human papillomavirus (hrHPV) types causes most invasive cervical cancers. Their oncogenic genome is encapsidated by L1 (major) and L2 (minor) coat proteins. Current HPV prophylactic vaccines are composed of L1 virus-like particles
Maninder Kaur et al.
Journal of food science and technology, 50(4), 809-814 (2014-01-16)
Effects of addition of different levels of gums (xanthan, carboxymethylcellulose and locust bean gum) on the pasting properties of tamarind kernel, potato and rice flour were studied by using Rapid Visco-Analyzer (RVA). Tamarind kernel powder (TKP) varied significantly (P < 0.05) from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service