Skip to Content
Merck
All Photos(2)

Key Documents

419044

Sigma-Aldrich

Cellulose acetate butyrate

average Mn ~70,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

mol wt

average Mn ~70,000

extent of labeling

12-15 wt. % Acetyl
35-39 wt. % Butyryl
1.2-2.2 wt. % Hydroxyl

refractive index

n20/D 1.475 (lit.)

density

1.25 g/mL at 25 °C (lit.)

SMILES string

O5[C@H]([C@@H]([C@H]([C@@H]([C@H]5COC(=O)CCC)OC(=O)CCC)OC(=O)CCC)OC(=O)CCC)O[C@@H]6[C@H](O[C@H]([C@@H]([C@H]6OC(=O)CCC)OC(=O)CCC)OC(=O)CCC)COC(=O)CCC.O3[C@H]([C@@H]([C@H]([C@@H]([C@H]3COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C)O[C@@H]4[C@H](O[C@H]([C@@H]([C@H]4OC(=

InChI key

WGKMZYZRLDSCLX-OLZTYWLQSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gheorghe Fundueanu et al.
Biomaterials, 26(20), 4337-4347 (2005-02-03)
Sulfopropylated dextran microspheres (SP-Ms), (Dm = 80 microm) loaded with a water soluble drug (Tetracycline HCl), were included in cellulose acetate butyrate (CAB) microcapsules. Spherical CAB microcapsules were obtained by oil in water (o/w) solvent evaporation method in the presence
Jun Wuk Park et al.
Macromolecular bioscience, 5(9), 840-852 (2005-09-02)
Miscible blends of PHB and CAB were prepared by the solvent-casting method with various blend compositions, and their orientation behavior was investigated during uniaxial drawing. X-ray analysis revealed that the orientation of the crystallizable PHB component in the drawn PHB/CAB
Soad A Yehia et al.
AAPS PharmSciTech, 10(1), 147-157 (2009-02-10)
The purpose of this study was to formulate budesonide (BUD) compression-coated tablets for colonic specific delivery. Pectin and guar gum were used as enzyme-dependent polymers. For comparison purposes, both pH- and time-dependent polymers were also tried. In vitro release studies
W M Obeidat et al.
Journal of microencapsulation, 21(1), 47-57 (2004-01-14)
Theophylline microspheres were prepared by the emulsion-solvent evaporation method using cellulose acetate butyrate (CAB381-20) and mixtures of CAB381-20(R) and cellulose acetate phthalate. The physical state of the drug, polymers and microspheres surfaces were determined using scanning electron microscopy. For those
Decheng Ma et al.
Journal of pharmaceutical and biomedical analysis, 35(4), 779-788 (2004-06-15)
The purpose of this study was to qualitatively and quantitatively determine potential cellulose acetate butyrate (CAB) extractables in a way to meaningfully predict the in vivo exposure resulting from clinical administration. Extractions of CAB-381-20 were performed in several solvent systems

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service