Skip to Content
Merck
All Photos(1)

Key Documents

415979

Sigma-Aldrich

2-(3-Bromopropoxy)tetrahydro-2H-pyran

98%

Synonym(s):

1-Bromo-3-(tetrahydropyranyloxy)propane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15BrO2
CAS Number:
Molecular Weight:
223.11
Beilstein:
106236
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

liquid

contains

potassium carbonate as stabilizer

refractive index

n20/D 1.478 (lit.)

bp

64.5 °C/0.8 mmHg (lit.)

density

1.317 g/mL at 25 °C (lit.)

functional group

bromo
ether

storage temp.

−20°C

SMILES string

BrCCCOC1CCCCO1

InChI

1S/C8H15BrO2/c9-5-3-7-11-8-4-1-2-6-10-8/h8H,1-7H2

InChI key

HJNHUFQGDJLQRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The latent alcohol functionality of this bifunctional building block can be generated using aqueous acid.[1][2]

Application

2-(3-Bromopropoxy)tetrahydro-2H-pyran may be used in organic synthesis studies.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The Journal of Organic Chemistry, 59, 2004-2004 (1994)
Tetrahedron Letters, 35, 4339-4339 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service