Recommended Products
Quality Level
Assay
98%
form
powder
mp
>300 °C (dec.) (lit.)
functional group
aldehyde
SMILES string
O=CC1=CNC(=O)NC1=O
InChI
1S/C5H4N2O3/c8-2-3-1-6-5(10)7-4(3)9/h1-2H,(H2,6,7,9,10)
InChI key
OHAMXGZMZZWRCA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
5-Formyluracil may be used for the preparation of covalently linked base with 5-aminocytosine pair via Schiff base formation.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Free radical biology & medicine, 47(4), 401-409 (2009-05-12)
The decomposition of organic hydroperoxides into peroxyl radicals is a potential source of singlet molecular oxygen [O2 (1Deltag)] in biological systems. This study shows that 5-(hydroperoxymethyl)uracil (5-HPMU), a thymine hydroperoxide within DNA, reacts with metal ions or HOCl, generating O2
Biochemistry, 43(10), 2682-2687 (2004-03-10)
5-Formyluracil (fU) is a major thymine lesion produced by reactive oxygen radicals and photosensitized oxidation. Although this residue is a potentially mutagenic lesion and is removed by several base excision repair enzymes, it is unknown whether fU is the substrate
Journal of the American Society for Mass Spectrometry, 13(10), 1190-1194 (2002-10-22)
2-Aminoimidazolone and 5-formyluracil are major one-electron photooxidation products of guanine and thymine in oligodeoxynucleotides (ODNs). Herein we report the HPLC isolation and tandem mass spectrometric characterization of ODNs carrying those types of base modifications. Collision-activated dissociation (CAD) of the deprotonated
Inorganic chemistry, 42(6), 2049-2055 (2003-03-18)
Two new 5-formyluracil thiosemicarbazone (H(3)ut) derivatives, Me-H(3)ut (1) and Me(2)-H(3)ut (2), were synthesized by reacting thiosemicarbazides, mono- and dimethylated on the aminic nitrogen, with 5-formyluracil and were subsequently characterized. These ligands, treated with copper chloride and nitrate, afforded three complexes:
Journal of the American Chemical Society, 127(47), 16681-16684 (2005-11-25)
We here present a novel covalently linked base pair via Schiff base formation between 5-formyluracil (fU) and 5-aminocytosine (AmC). Formation of the Schiff base linkage proceeds reversibly and does not require any additives. The cross-linked DNA is very stable under
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service